Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 138950222-138950495 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXL2NB
Entrez Gene ID
401089
Gene Name
FOXL2 neighbor
Gene Aliases
C3orf72
UniGene
Hs.591303
Gene Chromosome Location
Chr.3: 138947234-138953992 on Build GRCh38
Transcripts
NM_001040061.2 XM_005247443.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4935411 c.322G>A p.A108T Chr.3: 138950366-138950366 on Build GRCh38
4942433 c.339C>T p.L113L Chr.3: 138950383-138950383 on Build GRCh38
4771842 c.426C>T p.P142P Chr.3: 138950470-138950470 on Build GRCh38
4800529 c.378T>A p.V126V Chr.3: 138950422-138950422 on Build GRCh38
4667748 c.352G>A p.A118T Chr.3: 138950396-138950396 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748510602 Chr.3: 138950264-138950264 on Build GRCh38
rs760412852 Chr.3: 138950305-138950305 on Build GRCh38
rs369163230 Chr.3: 138950390-138950390 on Build GRCh38
rs775621751 Chr.3: 138950413-138950414 on Build GRCh38
rs771560242 Chr.3: 138950287-138950287 on Build GRCh38
rs761594173 Chr.3: 138950329-138950329 on Build GRCh38
rs769531270 Chr.3: 138950389-138950389 on Build GRCh38
rs762722486 Chr.3: 138950328-138950328 on Build GRCh38
rs775032717 Chr.3: 138950296-138950296 on Build GRCh38
rs112857156 Chr.3: 138950447-138950447 on Build GRCh38
rs752991018 Chr.3: 138950425-138950425 on Build GRCh38
rs766179348 Chr.3: 138950362-138950363 on Build GRCh38
rs778162603 Chr.3: 138950352-138950352 on Build GRCh38
rs1000923858 Chr.3: 138950319-138950319 on Build GRCh38
rs755308667 Chr.3: 138950342-138950342 on Build GRCh38
rs201934831 Chr.3: 138950371-138950371 on Build GRCh38
rs761612537 Chr.3: 138950383-138950383 on Build GRCh38
rs537914632 Chr.3: 138950474-138950474 on Build GRCh38
rs781560668 Chr.3: 138950343-138950343 on Build GRCh38
rs767863605 Chr.3: 138950405-138950405 on Build GRCh38
rs200299924 Chr.3: 138950253-138950253 on Build GRCh38
rs566392709 Chr.3: 138950404-138950404 on Build GRCh38
rs546443441 Chr.3: 138950396-138950396 on Build GRCh38
rs200544979 Chr.3: 138950430-138950430 on Build GRCh38
rs910540148 Chr.3: 138950471-138950471 on Build GRCh38
rs1024935963 Chr.3: 138950388-138950388 on Build GRCh38
rs1040636139 Chr.3: 138950246-138950246 on Build GRCh38
rs777257625 Chr.3: 138950269-138950269 on Build GRCh38
rs548632968 Chr.3: 138950340-138950340 on Build GRCh38
rs548433443 Chr.3: 138950327-138950327 on Build GRCh38
rs60650442 Chr.3: 138950380-138950380 on Build GRCh38
rs759620111 Chr.3: 138950402-138950402 on Build GRCh38
rs768099953 Chr.3: 138950458-138950458 on Build GRCh38
rs199657032 Chr.3: 138950475-138950475 on Build GRCh38
rs750318299 Chr.3: 138950332-138950332 on Build GRCh38
rs1042563968 Chr.3: 138950262-138950262 on Build GRCh38
rs530645680 Chr.3: 138950366-138950366 on Build GRCh38
rs902003770 Chr.3: 138950273-138950273 on Build GRCh38
rs199800109 Chr.3: 138950448-138950448 on Build GRCh38
rs773132889 Chr.3: 138950309-138950312 on Build GRCh38
rs764249769 Chr.3: 138950451-138950451 on Build GRCh38
rs754127471 Chr.3: 138950437-138950437 on Build GRCh38
rs770487946 Chr.3: 138950268-138950268 on Build GRCh38
rs745380004 Chr.3: 138950271-138950271 on Build GRCh38
rs1039079498 Chr.3: 138950331-138950331 on Build GRCh38
rs562194723 Chr.3: 138950338-138950338 on Build GRCh38
rs749760741 Chr.3: 138950353-138950353 on Build GRCh38
rs762966600 Chr.3: 138950337-138950337 on Build GRCh38
rs959383656 Chr.3: 138950463-138950463 on Build GRCh38
rs201515778 Chr.3: 138950394-138950394 on Build GRCh38
rs528480619 Chr.3: 138950304-138950304 on Build GRCh38
rs779660003 Chr.3: 138950370-138950370 on Build GRCh38
rs751562693 Chr.3: 138950339-138950339 on Build GRCh38
rs757845000 Chr.3: 138950445-138950445 on Build GRCh38
rs753114417 Chr.3: 138950345-138950345 on Build GRCh38
rs765166567 Chr.3: 138950330-138950330 on Build GRCh38
rs780846841 Chr.3: 138950460-138950460 on Build GRCh38
rs546476690 Chr.3: 138950250-138950250 on Build GRCh38
rs375623911 Chr.3: 138950424-138950424 on Build GRCh38
rs780154149 Chr.3: 138950243-138950243 on Build GRCh38
rs758648508 Chr.3: 138950443-138950443 on Build GRCh38
rs776473048 Chr.3: 138950318-138950318 on Build GRCh38
rs768279723 Chr.3: 138950306-138950306 on Build GRCh38

View Full Product Details