Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 86473332-86473573 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLCA1
Entrez Gene ID
1179
Gene Name
chloride channel accessory 1
Gene Aliases
CACC, CACC1, CLCRG1, CaCC-1, GOB5, hCLCA1, hCaCC-1
UniGene
Hs.194659
Gene Chromosome Location
Chr.1: 86468843-86500294 on Build GRCh38
Transcripts
NM_001285.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1602740 c.272A>T p.Y91F Chr.1: 86473526-86473526 on Build GRCh38
682840 c.212G>C p.R71P Chr.1: 86473466-86473466 on Build GRCh38
5911271 c.163-5C>T p.? Chr.1: 86473412-86473412 on Build GRCh38
4577623 c.166A>G p.M56V Chr.1: 86473420-86473420 on Build GRCh38
5381633 c.212G>A p.R71Q Chr.1: 86473466-86473466 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771331601 Chr.1: 86473362-86473362 on Build GRCh38
rs775277669 Chr.1: 86473431-86473431 on Build GRCh38
rs747575839 Chr.1: 86473405-86473405 on Build GRCh38
rs753031471 Chr.1: 86473506-86473506 on Build GRCh38
rs758680696 Chr.1: 86473509-86473509 on Build GRCh38
rs755172271 Chr.1: 86473473-86473473 on Build GRCh38
rs201967418 Chr.1: 86473394-86473394 on Build GRCh38
rs770200727 Chr.1: 86473476-86473476 on Build GRCh38
rs200401750 Chr.1: 86473387-86473387 on Build GRCh38
rs386633160 Chr.1: 86473480-86473482 on Build GRCh38
rs373779478 Chr.1: 86473527-86473527 on Build GRCh38
rs1006970208 Chr.1: 86473457-86473457 on Build GRCh38
rs180702059 Chr.1: 86473393-86473393 on Build GRCh38
rs753995315 Chr.1: 86473466-86473466 on Build GRCh38
rs769350528 Chr.1: 86473425-86473425 on Build GRCh38
rs761889678 Chr.1: 86473458-86473458 on Build GRCh38
rs373295226 Chr.1: 86473364-86473364 on Build GRCh38
rs912881855 Chr.1: 86473376-86473376 on Build GRCh38
rs774416063 Chr.1: 86473449-86473449 on Build GRCh38
rs779290264 Chr.1: 86473385-86473385 on Build GRCh38
rs200279591 Chr.1: 86473465-86473465 on Build GRCh38
rs1050696317 Chr.1: 86473439-86473439 on Build GRCh38
rs531139719 Chr.1: 86473520-86473520 on Build GRCh38
rs745490411 Chr.1: 86473422-86473422 on Build GRCh38
rs766535077 Chr.1: 86473461-86473461 on Build GRCh38
rs766372447 Chr.1: 86473534-86473534 on Build GRCh38
rs771786496 Chr.1: 86473420-86473420 on Build GRCh38
rs2145412 Chr.1: 86473447-86473447 on Build GRCh38
rs137892655 Chr.1: 86473434-86473434 on Build GRCh38
rs146279529 Chr.1: 86473380-86473380 on Build GRCh38
rs1015043218 Chr.1: 86473494-86473494 on Build GRCh38
rs778252130 Chr.1: 86473400-86473400 on Build GRCh38
rs370227290 Chr.1: 86473525-86473525 on Build GRCh38
rs367649206 Chr.1: 86473406-86473406 on Build GRCh38
rs765383297 Chr.1: 86473481-86473481 on Build GRCh38
rs757796481 Chr.1: 86473526-86473526 on Build GRCh38

View Full Product Details