Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 227016868-227017141 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC42BPA
Entrez Gene ID
8476
Gene Name
CDC42 binding protein kinase alpha
Gene Aliases
MRCK, MRCKA, PK428
UniGene
Hs.35433
Gene Chromosome Location
Chr.1: 226989865-227318492 on Build GRCh38
Transcripts
NM_003607.3 NM_014826.4 XM_005273317.3 XM_005273318.3 XM_005273320.3 XM_005273321.3 XM_005273322.3 XM_005273324.3 XM_006711834.3 XM_006711835.3 XM_011544306.2 XM_011544307.2 XM_011544308.2 XM_011544309.2 XM_017002578.1 XM_017002579.1 XM_017002580.1 XM_017002581.1 XM_017002582.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5451238 c.4623C>T p.D1541D Chr.1: 227017043-227017043 on Build GRCh38
4948946 c.4588C>T p.R1530C Chr.1: 227016973-227016973 on Build GRCh38
6865726 c.4568G>C p.R1523T Chr.1: 227016993-227016993 on Build GRCh38
4576964 c.4586G>A p.R1529Q Chr.1: 227016975-227016975 on Build GRCh38
5451237 c.4518C>T p.D1506D Chr.1: 227017043-227017043 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs571684122 Chr.1: 227017008-227017008 on Build GRCh38
rs371006782 Chr.1: 227016969-227016969 on Build GRCh38
rs575138409 Chr.1: 227017083-227017083 on Build GRCh38
rs770921984 Chr.1: 227016902-227016902 on Build GRCh38
rs763820783 Chr.1: 227016919-227016919 on Build GRCh38
rs555542592 Chr.1: 227016952-227016952 on Build GRCh38
rs747126532 Chr.1: 227016940-227016940 on Build GRCh38
rs756820444 Chr.1: 227016953-227016953 on Build GRCh38
rs774076384 Chr.1: 227017055-227017055 on Build GRCh38
rs956759499 Chr.1: 227017104-227017104 on Build GRCh38
rs780611349 Chr.1: 227016968-227016968 on Build GRCh38
rs368398399 Chr.1: 227017093-227017093 on Build GRCh38
rs201852267 Chr.1: 227016972-227016972 on Build GRCh38
rs146224129 Chr.1: 227017094-227017094 on Build GRCh38
rs755208656 Chr.1: 227017042-227017042 on Build GRCh38
rs752428735 Chr.1: 227016913-227016913 on Build GRCh38
rs754398931 Chr.1: 227017073-227017073 on Build GRCh38
rs374622112 Chr.1: 227016909-227016909 on Build GRCh38
rs368220176 Chr.1: 227016954-227016954 on Build GRCh38
rs767964615 Chr.1: 227016908-227016908 on Build GRCh38
rs555844878 Chr.1: 227017043-227017043 on Build GRCh38
rs745755276 Chr.1: 227017022-227017022 on Build GRCh38
rs763902584 Chr.1: 227017054-227017054 on Build GRCh38
rs544705386 Chr.1: 227017086-227017086 on Build GRCh38
rs368082253 Chr.1: 227016911-227016911 on Build GRCh38
rs776653752 Chr.1: 227017009-227017009 on Build GRCh38
rs762737464 Chr.1: 227016918-227016918 on Build GRCh38
rs761418365 Chr.1: 227017060-227017060 on Build GRCh38
rs751176656 Chr.1: 227016920-227016920 on Build GRCh38
rs1045581683 Chr.1: 227016976-227016976 on Build GRCh38
rs747096742 Chr.1: 227016990-227016990 on Build GRCh38
rs764892682 Chr.1: 227016912-227016912 on Build GRCh38
rs748479226 Chr.1: 227016974-227016974 on Build GRCh38
rs772328155 Chr.1: 227016981-227016981 on Build GRCh38
rs779106707 Chr.1: 227016973-227016973 on Build GRCh38
rs762832639 Chr.1: 227017045-227017045 on Build GRCh38
rs573904280 Chr.1: 227017072-227017072 on Build GRCh38

View Full Product Details