Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 173932375-173932612 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPEB4
Entrez Gene ID
80315
Gene Name
cytoplasmic polyadenylation element binding protein 4
Gene Aliases
CPE-BP4, hCPEB-4
UniGene
Hs.127126
Gene Chromosome Location
Chr.5: 173888328-173960991 on Build GRCh38
Transcripts
NM_001308189.1 NM_001308191.1 NM_001308192.1 NM_001308193.1 NM_030627.3 XM_005265994.1 XM_011534660.2 XM_011534661.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7125562 c.1208G>T p.G403V Chr.5: 173932450-173932450 on Build GRCh38
3853784 c.1211G>A p.R404H Chr.5: 173932453-173932453 on Build GRCh38
6634563 c.1254T>A p.I418I Chr.5: 173932496-173932496 on Build GRCh38
6170620 c.1252A>G p.I418V Chr.5: 173932494-173932494 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs781388011 Chr.5: 173932448-173932448 on Build GRCh38
rs952212300 Chr.5: 173932576-173932576 on Build GRCh38
rs951730414 Chr.5: 173932423-173932423 on Build GRCh38
rs111254945 Chr.5: 173932552-173932552 on Build GRCh38
rs1015341627 Chr.5: 173932462-173932462 on Build GRCh38
rs201191130 Chr.5: 173932534-173932534 on Build GRCh38
rs186178100 Chr.5: 173932426-173932426 on Build GRCh38
rs771165558 Chr.5: 173932407-173932407 on Build GRCh38
rs759833666 Chr.5: 173932409-173932409 on Build GRCh38
rs370585608 Chr.5: 173932422-173932422 on Build GRCh38
rs142106976 Chr.5: 173932453-173932453 on Build GRCh38
rs531165752 Chr.5: 173932431-173932431 on Build GRCh38
rs771413467 Chr.5: 173932517-173932517 on Build GRCh38
rs200019841 Chr.5: 173932478-173932478 on Build GRCh38
rs374648720 Chr.5: 173932548-173932548 on Build GRCh38
rs774733495 Chr.5: 173932433-173932433 on Build GRCh38
rs760248215 Chr.5: 173932414-173932414 on Build GRCh38
rs760756299 Chr.5: 173932477-173932477 on Build GRCh38
rs749355575 Chr.5: 173932479-173932479 on Build GRCh38
rs752970702 Chr.5: 173932452-173932452 on Build GRCh38
rs13187562 Chr.5: 173932558-173932558 on Build GRCh38
rs199814116 Chr.5: 173932408-173932408 on Build GRCh38
rs201491896 Chr.5: 173932412-173932412 on Build GRCh38
rs779632512 Chr.5: 173932542-173932542 on Build GRCh38
rs753797873 Chr.5: 173932417-173932417 on Build GRCh38
rs771693222 Chr.5: 173932410-173932410 on Build GRCh38
rs376228091 Chr.5: 173932430-173932430 on Build GRCh38
rs763746862 Chr.5: 173932415-173932415 on Build GRCh38
rs779295470 Chr.5: 173932541-173932541 on Build GRCh38

View Full Product Details