Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 180430217-180430442 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACBD6
Entrez Gene ID
84320
Gene Name
acyl-CoA binding domain containing 6
Gene Aliases
-
UniGene
Hs.200051
Gene Chromosome Location
Chr.1: 180269663-180502967 on Build GRCh38
Transcripts
NM_032360.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3803059 c.406G>T p.E136* Chr.1: 180430241-180430241 on Build GRCh38
4025712 c.425G>A p.G142D Chr.1: 180430222-180430222 on Build GRCh38
3385488 c.430C>T p.P144S Chr.1: 180430217-180430217 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs753095277 Chr.1: 180430269-180430269 on Build GRCh38
rs769867281 Chr.1: 180430362-180430362 on Build GRCh38
rs754995871 Chr.1: 180430313-180430313 on Build GRCh38
rs755647390 Chr.1: 180430300-180430300 on Build GRCh38
rs772781496 Chr.1: 180430253-180430261 on Build GRCh38
rs1033639404 Chr.1: 180430333-180430333 on Build GRCh38
rs760100075 Chr.1: 180430261-180430261 on Build GRCh38
rs959447807 Chr.1: 180430339-180430339 on Build GRCh38
rs779622631 Chr.1: 180430304-180430304 on Build GRCh38
rs758983860 Chr.1: 180430304-180430304 on Build GRCh38
rs749937786 Chr.1: 180430294-180430294 on Build GRCh38
rs950654058 Chr.1: 180430336-180430336 on Build GRCh38
rs200104643 Chr.1: 180430268-180430268 on Build GRCh38
rs964542348 Chr.1: 180430369-180430369 on Build GRCh38
rs765970620 Chr.1: 180430276-180430276 on Build GRCh38
rs181784939 Chr.1: 180430276-180430276 on Build GRCh38
rs753662160 Chr.1: 180430312-180430312 on Build GRCh38
rs746109375 Chr.1: 180430347-180430347 on Build GRCh38
rs961093391 Chr.1: 180430390-180430390 on Build GRCh38
rs762625867 Chr.1: 180430275-180430276 on Build GRCh38
rs1010294516 Chr.1: 180430253-180430253 on Build GRCh38
rs1026323583 Chr.1: 180430324-180430324 on Build GRCh38
rs771460498 Chr.1: 180430259-180430259 on Build GRCh38
rs760723613 Chr.1: 180430251-180430251 on Build GRCh38
rs751085365 Chr.1: 180430286-180430287 on Build GRCh38
rs1023081216 Chr.1: 180430243-180430243 on Build GRCh38
rs779290688 Chr.1: 180430277-180430277 on Build GRCh38

View Full Product Details