Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 70278108-70278367 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNOT2
Entrez Gene ID
4848
Gene Name
CCR4-NOT transcription complex subunit 2
Gene Aliases
CDC36, HSPC131, NOT2, NOT2H
UniGene
Hs.133350
Gene Chromosome Location
Chr.12: 70242993-70354993 on Build GRCh38
Transcripts
NM_001199302.1 NM_001199303.1 NM_014515.5 NR_037615.1 XM_006719429.3 XM_006719430.3 XM_006719432.3 XM_006719433.3 XM_011538399.1 XM_011538400.2 XM_017019348.1 XM_017019349.1 XM_017019350.1 XM_017019351.1 XM_017019352.1 XM_017019353.1 XM_017019354.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4673187 c.12T>C p.T4T Chr.12: 70278238-70278238 on Build GRCh38
943096 c.38G>T p.R13I Chr.12: 70278264-70278264 on Build GRCh38
6631444 c.15T>C p.D5D Chr.12: 70278241-70278241 on Build GRCh38
1238544 c.10A>G p.T4A Chr.12: 70278236-70278236 on Build GRCh38
7327365 c.9G>T p.R3S Chr.12: 70278235-70278235 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs747074882 Chr.12: 70278237-70278237 on Build GRCh38
rs770266923 Chr.12: 70278299-70278299 on Build GRCh38
rs758508570 Chr.12: 70278231-70278231 on Build GRCh38
rs943741115 Chr.12: 70278196-70278196 on Build GRCh38
rs779969072 Chr.12: 70278226-70278226 on Build GRCh38
rs372740317 Chr.12: 70278281-70278281 on Build GRCh38
rs368642986 Chr.12: 70278200-70278200 on Build GRCh38
rs753770253 Chr.12: 70278325-70278325 on Build GRCh38
rs992659321 Chr.12: 70278140-70278140 on Build GRCh38
rs188984517 Chr.12: 70278148-70278148 on Build GRCh38
rs759139669 Chr.12: 70278185-70278185 on Build GRCh38
rs777224437 Chr.12: 70278278-70278278 on Build GRCh38
rs929314432 Chr.12: 70278149-70278149 on Build GRCh38
rs764022385 Chr.12: 70278314-70278314 on Build GRCh38
rs866355171 Chr.12: 70278234-70278234 on Build GRCh38
rs760173781 Chr.12: 70278313-70278313 on Build GRCh38
rs769070211 Chr.12: 70278277-70278277 on Build GRCh38
rs377163945 Chr.12: 70278301-70278301 on Build GRCh38
rs200347802 Chr.12: 70278221-70278221 on Build GRCh38
rs756979338 Chr.12: 70278205-70278205 on Build GRCh38
rs372027107 Chr.12: 70278335-70278335 on Build GRCh38
rs753647248 Chr.12: 70278201-70278201 on Build GRCh38
rs779174363 Chr.12: 70278220-70278220 on Build GRCh38
rs767156197 Chr.12: 70278306-70278306 on Build GRCh38
rs182207624 Chr.12: 70278166-70278166 on Build GRCh38
rs766913194 Chr.12: 70278199-70278199 on Build GRCh38
rs774883554 Chr.12: 70278307-70278307 on Build GRCh38
rs369177196 Chr.12: 70278302-70278302 on Build GRCh38
rs545004853 Chr.12: 70278180-70278180 on Build GRCh38

View Full Product Details