Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 9715102-9715365 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPNE9
Entrez Gene ID
151835
Gene Name
copine family member 9
Gene Aliases
-
UniGene
Hs.121335
Gene Chromosome Location
Chr.3: 9703807-9729908 on Build GRCh38
Transcripts
NM_001308388.1 NM_153635.2 XM_011533386.2 XM_011533388.2 XM_011533389.2 XM_017005747.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3916777 c.714C>T p.F238F Chr.3: 9715310-9715310 on Build GRCh38
5719958 c.702C>T p.F234F Chr.3: 9715298-9715298 on Build GRCh38
3916776 c.714C>T p.F238F Chr.3: 9715310-9715310 on Build GRCh38
3945755 c.715A>T p.T239S Chr.3: 9715311-9715311 on Build GRCh38
5719957 c.702C>T p.F234F Chr.3: 9715298-9715298 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs112688518 Chr.3: 9715173-9715173 on Build GRCh38
rs149276313 Chr.3: 9715157-9715157 on Build GRCh38
rs778658020 Chr.3: 9715293-9715293 on Build GRCh38
rs374650389 Chr.3: 9715323-9715323 on Build GRCh38
rs1051017935 Chr.3: 9715143-9715143 on Build GRCh38
rs908227670 Chr.3: 9715303-9715303 on Build GRCh38
rs769844455 Chr.3: 9715340-9715340 on Build GRCh38
rs999357369 Chr.3: 9715198-9715198 on Build GRCh38
rs750074640 Chr.3: 9715300-9715300 on Build GRCh38
rs201103405 Chr.3: 9715292-9715292 on Build GRCh38
rs968525098 Chr.3: 9715193-9715193 on Build GRCh38
rs748216978 Chr.3: 9715333-9715333 on Build GRCh38
rs201005471 Chr.3: 9715307-9715307 on Build GRCh38
rs375348468 Chr.3: 9715255-9715255 on Build GRCh38
rs760079653 Chr.3: 9715254-9715254 on Build GRCh38
rs1000981182 Chr.3: 9715212-9715212 on Build GRCh38
rs1052788597 Chr.3: 9715228-9715228 on Build GRCh38
rs1009568139 Chr.3: 9715159-9715159 on Build GRCh38
rs199752870 Chr.3: 9715272-9715272 on Build GRCh38
rs997139425 Chr.3: 9715140-9715140 on Build GRCh38
rs903720784 Chr.3: 9715182-9715182 on Build GRCh38
rs768579411 Chr.3: 9715315-9715315 on Build GRCh38
rs670289 Chr.3: 9715278-9715278 on Build GRCh38
rs182380 Chr.3: 9715151-9715151 on Build GRCh38
rs144469531 Chr.3: 9715226-9715226 on Build GRCh38
rs753325896 Chr.3: 9715279-9715279 on Build GRCh38
rs559707497 Chr.3: 9715142-9715142 on Build GRCh38
rs201802299 Chr.3: 9715313-9715313 on Build GRCh38
rs183717410 Chr.3: 9715251-9715251 on Build GRCh38
rs779986468 Chr.3: 9715311-9715311 on Build GRCh38
rs773902470 Chr.3: 9715239-9715239 on Build GRCh38
rs372394053 Chr.3: 9715252-9715252 on Build GRCh38
rs773591671 Chr.3: 9715247-9715247 on Build GRCh38

View Full Product Details