Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 8373662-8373935 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANGPTL4
Entrez Gene ID
51129
Gene Name
angiopoietin like 4
Gene Aliases
ARP4, FIAF, HARP, HFARP, NL2, PGAR, TGQTL, UNQ171, pp1158
UniGene
Hs.9613
Gene Chromosome Location
Chr.19: 8364127-8374375 on Build GRCh38
Transcripts
NM_001039667.2 NM_139314.2 NR_104213.1 XM_005272484.3 XM_005272485.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3363087 c.1134C>G p.I378M Chr.19: 8373799-8373799 on Build GRCh38
7084713 c.1125G>A p.K375K Chr.19: 8373790-8373790 on Build GRCh38
3797714 c.1070C>G p.S357C Chr.19: 8373735-8373735 on Build GRCh38
6408545 c.1174G>A p.A392T Chr.19: 8373839-8373839 on Build GRCh38
6411880 c.1049G>A p.W350* Chr.19: 8373714-8373714 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776398253 Chr.19: 8373762-8373762 on Build GRCh38
rs1033145581 Chr.19: 8373712-8373712 on Build GRCh38
rs767768173 Chr.19: 8373860-8373860 on Build GRCh38
rs3183953 Chr.19: 8373745-8373745 on Build GRCh38
rs757722789 Chr.19: 8373808-8373808 on Build GRCh38
rs3210986 Chr.19: 8373889-8373889 on Build GRCh38
rs746836957 Chr.19: 8373821-8373821 on Build GRCh38
rs150107136 Chr.19: 8373758-8373758 on Build GRCh38
rs200103926 Chr.19: 8373839-8373839 on Build GRCh38
rs905582682 Chr.19: 8373912-8373912 on Build GRCh38
rs3210983 Chr.19: 8373879-8373879 on Build GRCh38
rs756440132 Chr.19: 8373780-8373780 on Build GRCh38
rs112807735 Chr.19: 8373888-8373888 on Build GRCh38
rs138460722 Chr.19: 8373759-8373759 on Build GRCh38
rs145509749 Chr.19: 8373887-8373887 on Build GRCh38
rs754161866 Chr.19: 8373882-8373882 on Build GRCh38
rs112739024 Chr.19: 8373703-8373703 on Build GRCh38
rs760839946 Chr.19: 8373870-8373870 on Build GRCh38
rs3210980 Chr.19: 8373871-8373871 on Build GRCh38
rs771783684 Chr.19: 8373752-8373752 on Build GRCh38
rs3210988 Chr.19: 8373892-8373892 on Build GRCh38
rs755264528 Chr.19: 8373772-8373772 on Build GRCh38
rs61512436 Chr.19: 8373894-8373894 on Build GRCh38
rs3210990 Chr.19: 8373901-8373901 on Build GRCh38
rs757767253 Chr.19: 8373885-8373885 on Build GRCh38
rs3210987 Chr.19: 8373891-8373891 on Build GRCh38
rs775257978 Chr.19: 8373819-8373819 on Build GRCh38
rs776431827 Chr.19: 8373828-8373828 on Build GRCh38
rs768365112 Chr.19: 8373826-8373826 on Build GRCh38
rs761768072 Chr.19: 8373765-8373765 on Build GRCh38
rs766384776 Chr.19: 8373770-8373770 on Build GRCh38
rs761664225 Chr.19: 8373830-8373830 on Build GRCh38
rs752950532 Chr.19: 8373867-8373867 on Build GRCh38
rs767717361 Chr.19: 8373776-8373776 on Build GRCh38
rs915909578 Chr.19: 8373805-8373805 on Build GRCh38
rs758382862 Chr.19: 8373687-8373687 on Build GRCh38
rs369070552 Chr.19: 8373754-8373754 on Build GRCh38
rs751594959 Chr.19: 8373771-8373771 on Build GRCh38
rs980207805 Chr.19: 8373700-8373700 on Build GRCh38
rs3210982 Chr.19: 8373875-8373875 on Build GRCh38
rs781414076 Chr.19: 8373878-8373879 on Build GRCh38
rs11672433 Chr.19: 8373832-8373832 on Build GRCh38
rs756672782 Chr.19: 8373706-8373706 on Build GRCh38
rs3210989 Chr.19: 8373896-8373896 on Build GRCh38
rs748871006 Chr.19: 8373727-8373727 on Build GRCh38
rs751754980 Chr.19: 8373689-8373689 on Build GRCh38
rs151099448 Chr.19: 8373715-8373715 on Build GRCh38
rs778289533 Chr.19: 8373813-8373813 on Build GRCh38
rs779669546 Chr.19: 8373904-8373904 on Build GRCh38
rs771904358 Chr.19: 8373816-8373816 on Build GRCh38
rs141084256 Chr.19: 8373796-8373796 on Build GRCh38
rs932056244 Chr.19: 8373787-8373787 on Build GRCh38
rs146942305 Chr.19: 8373815-8373815 on Build GRCh38
rs563408134 Chr.19: 8373688-8373688 on Build GRCh38
rs757615320 Chr.19: 8373744-8373745 on Build GRCh38
rs3210984 Chr.19: 8373881-8373881 on Build GRCh38
rs3210985 Chr.19: 8373883-8373883 on Build GRCh38
rs529369646 Chr.19: 8373699-8373699 on Build GRCh38
rs779488814 Chr.19: 8373777-8373777 on Build GRCh38
rs1010576800 Chr.19: 8373898-8373898 on Build GRCh38
rs755737249 Chr.19: 8373746-8373746 on Build GRCh38
rs778161135 Chr.19: 8373791-8373791 on Build GRCh38
rs769769905 Chr.19: 8373831-8373831 on Build GRCh38
rs3210981 Chr.19: 8373874-8373874 on Build GRCh38

View Full Product Details