Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 35675686-35675931 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CA9
Entrez Gene ID
768
Gene Name
carbonic anhydrase 9
Gene Aliases
CAIX, MN
UniGene
Hs.63287
Gene Chromosome Location
Chr.9: 35673859-35681159 on Build GRCh38
Transcripts
NM_001216.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6390321 c.478T>C p.F160L Chr.9: 35675805-35675805 on Build GRCh38
6368826 c.458C>A p.S153Y Chr.9: 35675785-35675785 on Build GRCh38
5461342 c.528C>G p.A176A Chr.9: 35675855-35675855 on Build GRCh38
2774378 c.505C>T p.Q169* Chr.9: 35675832-35675832 on Build GRCh38
1187716 c.439C>A p.P147T Chr.9: 35675766-35675766 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778853482 Chr.9: 35675860-35675860 on Build GRCh38
rs779568213 Chr.9: 35675750-35675750 on Build GRCh38
rs753162582 Chr.9: 35675722-35675722 on Build GRCh38
rs777909060 Chr.9: 35675781-35675781 on Build GRCh38
rs765821136 Chr.9: 35675834-35675834 on Build GRCh38
rs775685259 Chr.9: 35675812-35675812 on Build GRCh38
rs762307874 Chr.9: 35675826-35675826 on Build GRCh38
rs777976736 Chr.9: 35675845-35675846 on Build GRCh38
rs748045414 Chr.9: 35675775-35675775 on Build GRCh38
rs906641413 Chr.9: 35675889-35675889 on Build GRCh38
rs184826298 Chr.9: 35675802-35675802 on Build GRCh38
rs754676647 Chr.9: 35675760-35675760 on Build GRCh38
rs764623746 Chr.9: 35675742-35675742 on Build GRCh38
rs757229045 Chr.9: 35675898-35675898 on Build GRCh38
rs764462068 Chr.9: 35675823-35675823 on Build GRCh38
rs747271938 Chr.9: 35675867-35675867 on Build GRCh38
rs776990118 Chr.9: 35675874-35675874 on Build GRCh38
rs201849979 Chr.9: 35675716-35675716 on Build GRCh38
rs564223374 Chr.9: 35675818-35675818 on Build GRCh38
rs368797381 Chr.9: 35675808-35675808 on Build GRCh38
rs751102523 Chr.9: 35675756-35675756 on Build GRCh38
rs753767431 Chr.9: 35675803-35675804 on Build GRCh38
rs1024738924 Chr.9: 35675891-35675891 on Build GRCh38
rs776635903 Chr.9: 35675840-35675840 on Build GRCh38
rs754585567 Chr.9: 35675835-35675835 on Build GRCh38
rs761081719 Chr.9: 35675738-35675738 on Build GRCh38
rs773705392 Chr.9: 35675886-35675886 on Build GRCh38
rs779858764 Chr.9: 35675737-35675737 on Build GRCh38
rs866801817 Chr.9: 35675878-35675878 on Build GRCh38
rs759663992 Chr.9: 35675804-35675804 on Build GRCh38
rs377693403 Chr.9: 35675894-35675894 on Build GRCh38
rs777743411 Chr.9: 35675850-35675850 on Build GRCh38
rs762299422 Chr.9: 35675877-35675877 on Build GRCh38
rs1008355765 Chr.9: 35675858-35675858 on Build GRCh38
rs749261556 Chr.9: 35675790-35675790 on Build GRCh38
rs745812977 Chr.9: 35675913-35675913 on Build GRCh38
rs569356912 Chr.9: 35675870-35675870 on Build GRCh38
rs985781332 Chr.9: 35675709-35675709 on Build GRCh38
rs770643543 Chr.9: 35675773-35675773 on Build GRCh38
rs766960501 Chr.9: 35675888-35675888 on Build GRCh38
rs749910097 Chr.9: 35675743-35675743 on Build GRCh38
rs769826532 Chr.9: 35675779-35675779 on Build GRCh38
rs755926247 Chr.9: 35675849-35675849 on Build GRCh38
rs754300207 Chr.9: 35675825-35675825 on Build GRCh38
rs758429899 Chr.9: 35675833-35675833 on Build GRCh38
rs778957992 Chr.9: 35675907-35675907 on Build GRCh38
rs747260316 Chr.9: 35675865-35675865 on Build GRCh38
rs757322247 Chr.9: 35675859-35675859 on Build GRCh38
rs753752724 Chr.9: 35675897-35675897 on Build GRCh38
rs759137577 Chr.9: 35675887-35675887 on Build GRCh38
rs2301370 Chr.9: 35675855-35675855 on Build GRCh38
rs775729791 Chr.9: 35675863-35675863 on Build GRCh38
rs199516419 Chr.9: 35675868-35675868 on Build GRCh38
rs774667389 Chr.9: 35675714-35675714 on Build GRCh38
rs745998735 Chr.9: 35675861-35675861 on Build GRCh38
rs767716679 Chr.9: 35675721-35675721 on Build GRCh38
rs767242031 Chr.9: 35675839-35675839 on Build GRCh38
rs774494489 Chr.9: 35675803-35675803 on Build GRCh38
rs757851981 Chr.9: 35675746-35675746 on Build GRCh38
rs765842549 Chr.9: 35675882-35675882 on Build GRCh38

View Full Product Details