Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 148868822-148869095 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPA3
Entrez Gene ID
1359
Gene Name
carboxypeptidase A3
Gene Aliases
MC-CPA
UniGene
Hs.646
Gene Chromosome Location
Chr.3: 148865256-148897091 on Build GRCh38
Transcripts
NM_001870.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6096381 c.212G>A p.S71N Chr.3: 148868982-148868982 on Build GRCh38
6511639 c.146T>A p.L49H Chr.3: 148868916-148868916 on Build GRCh38
6096382 c.148G>C p.D50H Chr.3: 148868918-148868918 on Build GRCh38
310295 c.167C>A p.A56D Chr.3: 148868937-148868937 on Build GRCh38
1039671 c.187A>G p.N63D Chr.3: 148868957-148868957 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780164300 Chr.3: 148869012-148869012 on Build GRCh38
rs768095550 Chr.3: 148869070-148869070 on Build GRCh38
rs757071231 Chr.3: 148868934-148868934 on Build GRCh38
rs780045131 Chr.3: 148868945-148868945 on Build GRCh38
rs138685459 Chr.3: 148868948-148868948 on Build GRCh38
rs762108221 Chr.3: 148868899-148868899 on Build GRCh38
rs778216571 Chr.3: 148869052-148869052 on Build GRCh38
rs575898817 Chr.3: 148868999-148868999 on Build GRCh38
rs267599641 Chr.3: 148868990-148868990 on Build GRCh38
rs771798298 Chr.3: 148869058-148869058 on Build GRCh38
rs75478109 Chr.3: 148868955-148868955 on Build GRCh38
rs760123667 Chr.3: 148868974-148868974 on Build GRCh38
rs17850953 Chr.3: 148868959-148868959 on Build GRCh38
rs772174798 Chr.3: 148868870-148868870 on Build GRCh38
rs748661593 Chr.3: 148869034-148869034 on Build GRCh38
rs774219245 Chr.3: 148868972-148868972 on Build GRCh38
rs114531478 Chr.3: 148869016-148869016 on Build GRCh38
rs748515344 Chr.3: 148868954-148868954 on Build GRCh38
rs779071784 Chr.3: 148868936-148868936 on Build GRCh38
rs79854948 Chr.3: 148868889-148868889 on Build GRCh38
rs770277585 Chr.3: 148869043-148869043 on Build GRCh38
rs750227213 Chr.3: 148869008-148869008 on Build GRCh38
rs781709131 Chr.3: 148869030-148869030 on Build GRCh38
rs936553108 Chr.3: 148869032-148869032 on Build GRCh38
rs971432697 Chr.3: 148868854-148868854 on Build GRCh38
rs767194342 Chr.3: 148868911-148868911 on Build GRCh38
rs753756118 Chr.3: 148868976-148868976 on Build GRCh38
rs201601702 Chr.3: 148868901-148868901 on Build GRCh38
rs371537571 Chr.3: 148868907-148868907 on Build GRCh38
rs943735091 Chr.3: 148869007-148869007 on Build GRCh38
rs756011183 Chr.3: 148868921-148868921 on Build GRCh38
rs745904284 Chr.3: 148868865-148868865 on Build GRCh38
rs767282280 Chr.3: 148868971-148868971 on Build GRCh38
rs760198169 Chr.3: 148869067-148869067 on Build GRCh38
rs549388336 Chr.3: 148868853-148868853 on Build GRCh38
rs376722929 Chr.3: 148868932-148868932 on Build GRCh38
rs146101762 Chr.3: 148868947-148868947 on Build GRCh38
rs2270523 Chr.3: 148869011-148869011 on Build GRCh38
rs981564836 Chr.3: 148868882-148868882 on Build GRCh38
rs755054597 Chr.3: 148869017-148869017 on Build GRCh38
rs763949968 Chr.3: 148868927-148868927 on Build GRCh38
rs764049448 Chr.3: 148868975-148868975 on Build GRCh38
rs757019989 Chr.3: 148868978-148868978 on Build GRCh38
rs766091538 Chr.3: 148868900-148868900 on Build GRCh38
rs377599435 Chr.3: 148869056-148869056 on Build GRCh38
rs776998279 Chr.3: 148868897-148868897 on Build GRCh38
rs368902287 Chr.3: 148868892-148868892 on Build GRCh38
rs777037736 Chr.3: 148868952-148868952 on Build GRCh38
rs775966504 Chr.3: 148868877-148868877 on Build GRCh38
rs759327749 Chr.3: 148868960-148868960 on Build GRCh38
rs775032935 Chr.3: 148869060-148869060 on Build GRCh38
rs142834901 Chr.3: 148868939-148868939 on Build GRCh38
rs752571237 Chr.3: 148868916-148868916 on Build GRCh38

View Full Product Details