Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 57887750-57888003 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNGB1
Entrez Gene ID
1258
Gene Name
cyclic nucleotide gated channel beta 1
Gene Aliases
CNCG2, CNCG3L, CNCG4, CNG4, CNGB1B, GAR1, GARP, GARP2, RCNC2, RCNCb, RCNCbeta, RP45
UniGene
Hs.147062
Gene Chromosome Location
Chr.16: 57882340-57971116 on Build GRCh38
Transcripts
NM_001286130.1 NM_001297.4 XM_011522870.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
291862 c.3420C>T p.A1140A Chr.16: 57887897-57887897 on Build GRCh38
6273123 c.3422C>A p.A1141E Chr.16: 57887895-57887895 on Build GRCh38
2992490 c.3330C>T p.N1110N Chr.16: 57887987-57887987 on Build GRCh38
3818167 c.3360T>G p.G1120G Chr.16: 57887957-57887957 on Build GRCh38
971904 c.3331G>A p.A1111T Chr.16: 57887986-57887986 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756212388 Chr.16: 57887869-57887869 on Build GRCh38
rs748808412 Chr.16: 57887978-57887978 on Build GRCh38
rs774214610 Chr.16: 57887898-57887898 on Build GRCh38
rs868052603 Chr.16: 57887968-57887968 on Build GRCh38
rs778177540 Chr.16: 57887948-57887948 on Build GRCh38
rs1054960787 Chr.16: 57887804-57887804 on Build GRCh38
rs368962905 Chr.16: 57887847-57887847 on Build GRCh38
rs760925048 Chr.16: 57887910-57887910 on Build GRCh38
rs981771347 Chr.16: 57887933-57887933 on Build GRCh38
rs375525176 Chr.16: 57887887-57887887 on Build GRCh38
rs745504458 Chr.16: 57887894-57887894 on Build GRCh38
rs771473576 Chr.16: 57887892-57887892 on Build GRCh38
rs755577473 Chr.16: 57887885-57887885 on Build GRCh38
rs202011905 Chr.16: 57887822-57887822 on Build GRCh38
rs753974570 Chr.16: 57887914-57887914 on Build GRCh38
rs768138983 Chr.16: 57887985-57887985 on Build GRCh38
rs369943028 Chr.16: 57887920-57887920 on Build GRCh38
rs761651354 Chr.16: 57887901-57887901 on Build GRCh38
rs564430138 Chr.16: 57887827-57887827 on Build GRCh38
rs745429125 Chr.16: 57887805-57887805 on Build GRCh38
rs751822516 Chr.16: 57887951-57887951 on Build GRCh38
rs11076207 Chr.16: 57887848-57887848 on Build GRCh38
rs760675556 Chr.16: 57887780-57887780 on Build GRCh38
rs200242407 Chr.16: 57887977-57887977 on Build GRCh38
rs774886960 Chr.16: 57887940-57887949 on Build GRCh38
rs769343878 Chr.16: 57887810-57887810 on Build GRCh38
rs763970905 Chr.16: 57887839-57887839 on Build GRCh38
rs755822581 Chr.16: 57887976-57887976 on Build GRCh38
rs934642131 Chr.16: 57887792-57887792 on Build GRCh38
rs754441053 Chr.16: 57887873-57887873 on Build GRCh38
rs11867123 Chr.16: 57887939-57887939 on Build GRCh38
rs1017713632 Chr.16: 57887836-57887836 on Build GRCh38
rs753253429 Chr.16: 57887931-57887931 on Build GRCh38
rs201449358 Chr.16: 57887896-57887896 on Build GRCh38
rs750641757 Chr.16: 57887853-57887853 on Build GRCh38
rs376171765 Chr.16: 57887902-57887902 on Build GRCh38
rs370962776 Chr.16: 57887886-57887886 on Build GRCh38
rs562161300 Chr.16: 57887895-57887895 on Build GRCh38
rs371860152 Chr.16: 57887897-57887897 on Build GRCh38
rs202089209 Chr.16: 57887900-57887900 on Build GRCh38
rs375348116 Chr.16: 57887919-57887919 on Build GRCh38
rs766474948 Chr.16: 57887913-57887913 on Build GRCh38
rs373477921 Chr.16: 57887938-57887938 on Build GRCh38
rs893198760 Chr.16: 57887809-57887809 on Build GRCh38
rs540706326 Chr.16: 57887891-57887891 on Build GRCh38

View Full Product Details