Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 154284474-154284705 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHX36
Entrez Gene ID
170506
Gene Name
DEAH-box helicase 36
Gene Aliases
DDX36, G4R1, MLEL1, RHAU
UniGene
Hs.446270
Gene Chromosome Location
Chr.3: 154275668-154324497 on Build GRCh38
Transcripts
NM_001114397.1 NM_020865.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4114619 c.2174A>C p.K725T Chr.3: 154284659-154284659 on Build GRCh38
5457031 c.2286G>A p.A762A Chr.3: 154284589-154284589 on Build GRCh38
4114617 c.2224C>T p.H742Y Chr.3: 154284609-154284609 on Build GRCh38
5457032 c.2244G>A p.A748A Chr.3: 154284589-154284589 on Build GRCh38
1661443 c.2207G>A p.G736E Chr.3: 154284668-154284668 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs866934529 Chr.3: 154284561-154284561 on Build GRCh38
rs748525457 Chr.3: 154284589-154284589 on Build GRCh38
rs754654287 Chr.3: 154284568-154284568 on Build GRCh38
rs1050710113 Chr.3: 154284625-154284625 on Build GRCh38
rs765233520 Chr.3: 154284547-154284547 on Build GRCh38
rs1006752629 Chr.3: 154284662-154284662 on Build GRCh38
rs757932256 Chr.3: 154284550-154284550 on Build GRCh38
rs533904004 Chr.3: 154284566-154284566 on Build GRCh38
rs746732662 Chr.3: 154284659-154284660 on Build GRCh38
rs200630334 Chr.3: 154284578-154284578 on Build GRCh38
rs773671514 Chr.3: 154284596-154284596 on Build GRCh38
rs773260677 Chr.3: 154284590-154284590 on Build GRCh38
rs756752265 Chr.3: 154284569-154284569 on Build GRCh38
rs376802780 Chr.3: 154284619-154284619 on Build GRCh38
rs763254140 Chr.3: 154284624-154284624 on Build GRCh38
rs759657448 Chr.3: 154284509-154284509 on Build GRCh38
rs889193975 Chr.3: 154284647-154284647 on Build GRCh38
rs554438716 Chr.3: 154284568-154284569 on Build GRCh38
rs566541234 Chr.3: 154284549-154284549 on Build GRCh38
rs774480904 Chr.3: 154284630-154284630 on Build GRCh38
rs562366213 Chr.3: 154284578-154284579 on Build GRCh38
rs757048192 Chr.3: 154284535-154284535 on Build GRCh38
rs759590228 Chr.3: 154284648-154284648 on Build GRCh38
rs867369899 Chr.3: 154284581-154284582 on Build GRCh38
rs777342103 Chr.3: 154284564-154284565 on Build GRCh38
rs116767159 Chr.3: 154284533-154284533 on Build GRCh38
rs550537742 Chr.3: 154284541-154284541 on Build GRCh38
rs369705579 Chr.3: 154284522-154284522 on Build GRCh38
rs914280304 Chr.3: 154284526-154284526 on Build GRCh38
rs753400803 Chr.3: 154284567-154284568 on Build GRCh38
rs779722858 Chr.3: 154284552-154284552 on Build GRCh38
rs767754856 Chr.3: 154284658-154284658 on Build GRCh38
rs796697991 Chr.3: 154284569-154284570 on Build GRCh38
rs200020002 Chr.3: 154284569-154284570 on Build GRCh38
rs1030943706 Chr.3: 154284498-154284498 on Build GRCh38
rs969131416 Chr.3: 154284601-154284601 on Build GRCh38
rs780768054 Chr.3: 154284569-154284569 on Build GRCh38

View Full Product Details