Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 83008529-83008792 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C15orf40
Entrez Gene ID
123207
Gene Name
chromosome 15 open reading frame 40
Gene Aliases
-
UniGene
Hs.352541
Gene Chromosome Location
Chr.15: 82988297-83012298 on Build GRCh38
Transcripts
NM_001160113.1 NM_001160114.1 NM_001160115.1 NM_001160116.1 NM_144597.2 NR_027649.1 NR_027650.1 XM_006720385.2 XM_011521212.2 XM_011521213.2 XM_011521214.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5849928 c.233G>A p.R78Q Chr.15: 83008600-83008600 on Build GRCh38
3816897 c.345G>T p.K115N Chr.15: 83008569-83008569 on Build GRCh38
6327530 c.250C>T p.L84L Chr.15: 83008583-83008583 on Build GRCh38
6327531 c.331C>T p.L111L Chr.15: 83008583-83008583 on Build GRCh38
5054543 c.340A>G p.R114G Chr.15: 83008574-83008574 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs190463970 Chr.15: 83008715-83008715 on Build GRCh38
rs775771446 Chr.15: 83008701-83008701 on Build GRCh38
rs376937425 Chr.15: 83008649-83008649 on Build GRCh38
rs777407627 Chr.15: 83008654-83008654 on Build GRCh38
rs762359737 Chr.15: 83008708-83008710 on Build GRCh38
rs774650229 Chr.15: 83008708-83008708 on Build GRCh38
rs370343641 Chr.15: 83008705-83008705 on Build GRCh38
rs771275898 Chr.15: 83008562-83008562 on Build GRCh38
rs535124808 Chr.15: 83008607-83008607 on Build GRCh38
rs765392069 Chr.15: 83008580-83008580 on Build GRCh38
rs767407114 Chr.15: 83008623-83008623 on Build GRCh38
rs746428504 Chr.15: 83008680-83008680 on Build GRCh38
rs1006749061 Chr.15: 83008666-83008666 on Build GRCh38
rs554919886 Chr.15: 83008719-83008719 on Build GRCh38
rs750313793 Chr.15: 83008626-83008626 on Build GRCh38
rs758109349 Chr.15: 83008651-83008651 on Build GRCh38
rs755945726 Chr.15: 83008627-83008627 on Build GRCh38
rs72755956 Chr.15: 83008763-83008763 on Build GRCh38
rs1007517708 Chr.15: 83008737-83008737 on Build GRCh38
rs151282701 Chr.15: 83008673-83008673 on Build GRCh38
rs116091515 Chr.15: 83008566-83008566 on Build GRCh38
rs760340569 Chr.15: 83008711-83008711 on Build GRCh38
rs897835449 Chr.15: 83008572-83008572 on Build GRCh38
rs773620548 Chr.15: 83008554-83008554 on Build GRCh38
rs955371829 Chr.15: 83008619-83008619 on Build GRCh38
rs139727269 Chr.15: 83008734-83008734 on Build GRCh38
rs766172072 Chr.15: 83008637-83008637 on Build GRCh38
rs747542516 Chr.15: 83008560-83008560 on Build GRCh38
rs754412040 Chr.15: 83008642-83008642 on Build GRCh38
rs753838125 Chr.15: 83008640-83008640 on Build GRCh38
rs763317306 Chr.15: 83008618-83008618 on Build GRCh38
rs566813563 Chr.15: 83008601-83008601 on Build GRCh38
rs202223471 Chr.15: 83008647-83008647 on Build GRCh38
rs142320183 Chr.15: 83008620-83008620 on Build GRCh38
rs377371105 Chr.15: 83008675-83008675 on Build GRCh38
rs771347862 Chr.15: 83008679-83008679 on Build GRCh38
rs769973930 Chr.15: 83008688-83008688 on Build GRCh38
rs1030449901 Chr.15: 83008616-83008616 on Build GRCh38

View Full Product Details