Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 99500687-99500954 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNK
Entrez Gene ID
8812
Gene Name
cyclin K
Gene Aliases
CPR4
UniGene
Hs.510409
Gene Chromosome Location
Chr.14: 99481402-99511515 on Build GRCh38
Transcripts
NM_001099402.1 XM_005268154.4 XM_011537275.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7263914 c.476A>G p.H159R Chr.14: 99500830-99500830 on Build GRCh38
3999438 c.495A>T p.L165L Chr.14: 99500849-99500849 on Build GRCh38
7187675 c.493C>T p.L165L Chr.14: 99500847-99500847 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs183907313 Chr.14: 99500920-99500920 on Build GRCh38
rs776917591 Chr.14: 99500727-99500727 on Build GRCh38
rs868868559 Chr.14: 99500833-99500833 on Build GRCh38
rs143873475 Chr.14: 99500724-99500724 on Build GRCh38
rs201567868 Chr.14: 99500874-99500876 on Build GRCh38
rs1004054099 Chr.14: 99500763-99500763 on Build GRCh38
rs202052846 Chr.14: 99500855-99500855 on Build GRCh38
rs765350035 Chr.14: 99500746-99500746 on Build GRCh38
rs373287151 Chr.14: 99500717-99500717 on Build GRCh38
rs759677002 Chr.14: 99500737-99500737 on Build GRCh38
rs868057477 Chr.14: 99500814-99500814 on Build GRCh38
rs2069493 Chr.14: 99500849-99500849 on Build GRCh38
rs760419179 Chr.14: 99500898-99500898 on Build GRCh38
rs560030876 Chr.14: 99500906-99500906 on Build GRCh38
rs879170478 Chr.14: 99500837-99500837 on Build GRCh38
rs773413663 Chr.14: 99500861-99500861 on Build GRCh38
rs376813581 Chr.14: 99500729-99500729 on Build GRCh38
rs763071876 Chr.14: 99500922-99500922 on Build GRCh38
rs869192674 Chr.14: 99500884-99500884 on Build GRCh38
rs918156496 Chr.14: 99500878-99500878 on Build GRCh38
rs767370737 Chr.14: 99500853-99500853 on Build GRCh38
rs1026727837 Chr.14: 99500721-99500721 on Build GRCh38
rs774436537 Chr.14: 99500752-99500752 on Build GRCh38
rs750167304 Chr.14: 99500889-99500889 on Build GRCh38
rs148603851 Chr.14: 99500917-99500917 on Build GRCh38
rs375840284 Chr.14: 99500882-99500882 on Build GRCh38

View Full Product Details