Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 138157097-138157370 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRD8
Entrez Gene ID
10902
Gene Name
bromodomain containing 8
Gene Aliases
SMAP, SMAP2, p120
UniGene
Hs.519337
Gene Chromosome Location
Chr.5: 138139766-138178669 on Build GRCh38
Transcripts
NM_001164326.1 NM_001300961.1 NM_001300962.1 NM_001300966.1 NM_006696.3 NM_139199.1 XM_005271855.2 XM_005271856.2 XM_005271857.2 XM_005271859.3 XM_005271860.3 XM_005271861.3 XM_011543109.1 XM_011543110.1 XM_017008969.1 XM_017008970.1 XM_017008971.1 XM_017008972.1 XM_017008973.1 XM_017008974.1 XM_017008975.1 XM_017008976.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1594791 c.2819G>A p.R940H Chr.5: 138157245-138157245 on Build GRCh38
4419724 c.2797-6A>G p.? Chr.5: 138157273-138157273 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs140446934 Chr.5: 138157340-138157340 on Build GRCh38
rs986426264 Chr.5: 138157140-138157140 on Build GRCh38
rs765326085 Chr.5: 138157310-138157310 on Build GRCh38
rs765007676 Chr.5: 138157124-138157124 on Build GRCh38
rs777528061 Chr.5: 138157161-138157161 on Build GRCh38
rs781119248 Chr.5: 138157173-138157173 on Build GRCh38
rs979581251 Chr.5: 138157133-138157133 on Build GRCh38
rs1055765962 Chr.5: 138157337-138157337 on Build GRCh38
rs905973861 Chr.5: 138157323-138157323 on Build GRCh38
rs938054185 Chr.5: 138157336-138157336 on Build GRCh38
rs190873320 Chr.5: 138157338-138157338 on Build GRCh38
rs758885026 Chr.5: 138157204-138157204 on Build GRCh38
rs770898724 Chr.5: 138157172-138157172 on Build GRCh38
rs746933343 Chr.5: 138157165-138157165 on Build GRCh38
rs919096832 Chr.5: 138157226-138157226 on Build GRCh38
rs1002852873 Chr.5: 138157325-138157325 on Build GRCh38
rs370027543 Chr.5: 138157192-138157192 on Build GRCh38
rs535341507 Chr.5: 138157322-138157322 on Build GRCh38
rs548162258 Chr.5: 138157327-138157327 on Build GRCh38
rs201918313 Chr.5: 138157181-138157181 on Build GRCh38
rs761160856 Chr.5: 138157224-138157224 on Build GRCh38
rs754138737 Chr.5: 138157303-138157303 on Build GRCh38
rs758374567 Chr.5: 138157157-138157157 on Build GRCh38
rs375727517 Chr.5: 138157185-138157185 on Build GRCh38
rs753221957 Chr.5: 138157167-138157168 on Build GRCh38
rs1009483292 Chr.5: 138157348-138157348 on Build GRCh38
rs372362803 Chr.5: 138157141-138157141 on Build GRCh38
rs1042546567 Chr.5: 138157343-138157343 on Build GRCh38
rs78219553 Chr.5: 138157273-138157273 on Build GRCh38
rs957047188 Chr.5: 138157136-138157136 on Build GRCh38
rs151262871 Chr.5: 138157193-138157193 on Build GRCh38
rs372126634 Chr.5: 138157305-138157305 on Build GRCh38
rs750619996 Chr.5: 138157177-138157177 on Build GRCh38
rs762330652 Chr.5: 138157190-138157190 on Build GRCh38

View Full Product Details