Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155721466-155721695 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAP3
Entrez Gene ID
7818
Gene Name
death associated protein 3
Gene Aliases
DAP-3, MRP-S29, MRPS29, bMRP-10
UniGene
Hs.516746
Gene Chromosome Location
Chr.1: 155688104-155739010 on Build GRCh38
Transcripts
NM_001199849.1 NM_001199850.1 NM_001199851.1 NM_004632.3 NM_033657.2 XM_005245480.2 XM_005245481.2 XM_005245482.4 XM_011509972.2 XM_017002289.1 XM_017002290.1 XM_017002291.1 XM_017002292.1 XM_017002293.1 XM_017002294.1 XM_017002295.1 XM_017002296.1 XM_017002297.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5716154 c.213C>T p.S71S Chr.1: 155721561-155721561 on Build GRCh38
5701430 c.212_213insC p.Q73fs*24 Chr.1: 155721560-155721561 on Build GRCh38
6640000 c.268C>T p.Q90* Chr.1: 155721616-155721616 on Build GRCh38
5652919 c.170C>A p.A57D Chr.1: 155721518-155721518 on Build GRCh38
6484948 c.186G>C p.Q62H Chr.1: 155721534-155721534 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762857520 Chr.1: 155721667-155721667 on Build GRCh38
rs996259799 Chr.1: 155721665-155721665 on Build GRCh38
rs750118220 Chr.1: 155721500-155721500 on Build GRCh38
rs758124736 Chr.1: 155721565-155721565 on Build GRCh38
rs763570884 Chr.1: 155721542-155721542 on Build GRCh38
rs776111877 Chr.1: 155721633-155721633 on Build GRCh38
rs751431768 Chr.1: 155721586-155721586 on Build GRCh38
rs749493060 Chr.1: 155721501-155721501 on Build GRCh38
rs769983810 Chr.1: 155721495-155721495 on Build GRCh38
rs761138094 Chr.1: 155721636-155721636 on Build GRCh38
rs780098429 Chr.1: 155721571-155721571 on Build GRCh38
rs766255365 Chr.1: 155721615-155721615 on Build GRCh38
rs757230517 Chr.1: 155721559-155721560 on Build GRCh38
rs760288202 Chr.1: 155721537-155721537 on Build GRCh38
rs775853011 Chr.1: 155721530-155721530 on Build GRCh38
rs750282457 Chr.1: 155721662-155721663 on Build GRCh38
rs922535122 Chr.1: 155721576-155721576 on Build GRCh38
rs765000883 Chr.1: 155721560-155721560 on Build GRCh38
rs768185601 Chr.1: 155721638-155721638 on Build GRCh38
rs1049297931 Chr.1: 155721626-155721626 on Build GRCh38
rs748327569 Chr.1: 155721493-155721493 on Build GRCh38
rs112771778 Chr.1: 155721560-155721560 on Build GRCh38
rs540380 Chr.1: 155721644-155721644 on Build GRCh38
rs772308093 Chr.1: 155721628-155721628 on Build GRCh38
rs774733542 Chr.1: 155721522-155721522 on Build GRCh38
rs1802737 Chr.1: 155721595-155721595 on Build GRCh38
rs758197249 Chr.1: 155721670-155721671 on Build GRCh38
rs779146216 Chr.1: 155721611-155721611 on Build GRCh38
rs753684492 Chr.1: 155721493-155721493 on Build GRCh38
rs374151645 Chr.1: 155721590-155721590 on Build GRCh38
rs753527370 Chr.1: 155721554-155721554 on Build GRCh38
rs767996923 Chr.1: 155721529-155721529 on Build GRCh38
rs758451367 Chr.1: 155721669-155721669 on Build GRCh38
rs1005098261 Chr.1: 155721639-155721639 on Build GRCh38
rs902024769 Chr.1: 155721646-155721646 on Build GRCh38
rs187192135 Chr.1: 155721589-155721589 on Build GRCh38
rs750031258 Chr.1: 155721660-155721660 on Build GRCh38
rs777964128 Chr.1: 155721594-155721594 on Build GRCh38
rs750166849 Chr.1: 155721564-155721564 on Build GRCh38
rs570752872 Chr.1: 155721588-155721588 on Build GRCh38
rs573427333 Chr.1: 155721650-155721650 on Build GRCh38
rs759970366 Chr.1: 155721523-155721523 on Build GRCh38
rs771223930 Chr.1: 155721606-155721606 on Build GRCh38
rs879023821 Chr.1: 155721541-155721541 on Build GRCh38
rs749547998 Chr.1: 155721604-155721604 on Build GRCh38

View Full Product Details