Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 86970523-86970756 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADIRF
Entrez Gene ID
10974
Gene Name
adipogenesis regulatory factor
Gene Aliases
AFRO, APM2, C10orf116, apM-2
UniGene
Hs.642660
Gene Chromosome Location
Chr.10: 86968431-86970909 on Build GRCh38
Transcripts
NM_006829.2 NM_133447.1 XM_006717970.3

Gene Symbol
AGAP11
Entrez Gene ID
119385
Gene Name
ArfGAP with GTPase domain, ankyrin repeat and PH domain 11
Gene Aliases
-
UniGene
Hs.511787
Gene Chromosome Location
Chr.10: 86970741-87010203 on Build GRCh38
Transcripts
NM_006829.2 NM_133447.1 XM_006717970.3

Gene Symbol
MMRN2
Entrez Gene ID
79812
Gene Name
multimerin 2
Gene Aliases
EMILIN-3, EMILIN3, ENDOGLYX-1
UniGene
Hs.524479
Gene Chromosome Location
Chr.10: 86935540-86971311 on Build GRCh38
Transcripts
NM_006829.2 NM_133447.1 XM_006717970.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7206599 c.214T>A p.F72I Chr.10: 86970565-86970565 on Build GRCh38
5453778 c.189C>T p.D63D Chr.10: 86970540-86970540 on Build GRCh38
146987 c.204T>C p.I68I Chr.10: 86970555-86970555 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs372463086 Chr.10: 86970578-86970578 on Build GRCh38
rs756892263 Chr.10: 86970592-86970592 on Build GRCh38
rs1024272222 Chr.10: 86970680-86970680 on Build GRCh38
rs766525730 Chr.10: 86970616-86970616 on Build GRCh38
rs774588082 Chr.10: 86970696-86970696 on Build GRCh38
rs1804881 Chr.10: 86970666-86970666 on Build GRCh38
rs1012466421 Chr.10: 86970679-86970679 on Build GRCh38
rs750509133 Chr.10: 86970617-86970618 on Build GRCh38
rs775453803 Chr.10: 86970601-86970601 on Build GRCh38
rs745316875 Chr.10: 86970597-86970597 on Build GRCh38
rs112083892 Chr.10: 86970733-86970733 on Build GRCh38
rs781682872 Chr.10: 86970568-86970568 on Build GRCh38
rs756486380 Chr.10: 86970631-86970631 on Build GRCh38
rs754022420 Chr.10: 86970636-86970637 on Build GRCh38
rs368188117 Chr.10: 86970567-86970567 on Build GRCh38
rs750609216 Chr.10: 86970551-86970551 on Build GRCh38
rs768472913 Chr.10: 86970603-86970603 on Build GRCh38
rs767962669 Chr.10: 86970566-86970599 on Build GRCh38
rs7960 Chr.10: 86970617-86970617 on Build GRCh38
rs531379548 Chr.10: 86970642-86970642 on Build GRCh38
rs114065058 Chr.10: 86970632-86970632 on Build GRCh38
rs376371857 Chr.10: 86970602-86970602 on Build GRCh38
rs760678243 Chr.10: 86970622-86970622 on Build GRCh38
rs998593202 Chr.10: 86970719-86970719 on Build GRCh38
rs900347805 Chr.10: 86970600-86970600 on Build GRCh38
rs376853320 Chr.10: 86970556-86970556 on Build GRCh38
rs759612398 Chr.10: 86970619-86970619 on Build GRCh38
rs372989236 Chr.10: 86970669-86970669 on Build GRCh38
rs746956212 Chr.10: 86970705-86970705 on Build GRCh38
rs1049250 Chr.10: 86970620-86970620 on Build GRCh38
rs779901691 Chr.10: 86970685-86970685 on Build GRCh38
rs766457742 Chr.10: 86970633-86970634 on Build GRCh38
rs1804882 Chr.10: 86970558-86970558 on Build GRCh38
rs74503710 Chr.10: 86970652-86970652 on Build GRCh38
rs765406198 Chr.10: 86970549-86970549 on Build GRCh38
rs111937931 Chr.10: 86970732-86970732 on Build GRCh38
rs10653 Chr.10: 86970627-86970627 on Build GRCh38
rs1049241 Chr.10: 86970586-86970586 on Build GRCh38
rs755236211 Chr.10: 86970650-86970650 on Build GRCh38
rs894359753 Chr.10: 86970639-86970639 on Build GRCh38
rs111880726 Chr.10: 86970662-86970662 on Build GRCh38
rs752130957 Chr.10: 86970563-86970563 on Build GRCh38
rs754187752 Chr.10: 86970637-86970637 on Build GRCh38
rs778570339 Chr.10: 86970596-86970596 on Build GRCh38
rs4869 Chr.10: 86970555-86970555 on Build GRCh38
rs748458433 Chr.10: 86970706-86970706 on Build GRCh38
rs756463479 Chr.10: 86970576-86970576 on Build GRCh38
rs767941860 Chr.10: 86970622-86970622 on Build GRCh38
rs14656 Chr.10: 86970629-86970629 on Build GRCh38
rs886179760 Chr.10: 86970625-86970625 on Build GRCh38
rs757892121 Chr.10: 86970657-86970657 on Build GRCh38
rs199985594 Chr.10: 86970598-86970598 on Build GRCh38
rs1036428663 Chr.10: 86970610-86970610 on Build GRCh38
rs375966291 Chr.10: 86970607-86970607 on Build GRCh38
rs1038948443 Chr.10: 86970589-86970589 on Build GRCh38
rs763160162 Chr.10: 86970608-86970608 on Build GRCh38

View Full Product Details