Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 73816633-73816872 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2orf78
Entrez Gene ID
388960
Gene Name
chromosome 2 open reading frame 78
Gene Aliases
COG5373, hCG1989538
UniGene
Hs.456833
Gene Chromosome Location
Chr.2: 73784189-73817148 on Build GRCh38
Transcripts
NM_001080474.1 XM_005264302.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4481672 c.2530C>T p.Q844* Chr.2: 73816753-73816753 on Build GRCh38
3840012 c.2372C>T p.S791F Chr.2: 73816685-73816685 on Build GRCh38
5032090 c.2636G>A p.R879H Chr.2: 73816859-73816859 on Build GRCh38
5803608 c.2551A>T p.S851C Chr.2: 73816774-73816774 on Build GRCh38
5736090 c.2471C>T p.P824L Chr.2: 73816784-73816784 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs934293521 Chr.2: 73816729-73816729 on Build GRCh38
rs750694636 Chr.2: 73816830-73816830 on Build GRCh38
rs781351281 Chr.2: 73816658-73816658 on Build GRCh38
rs534131690 Chr.2: 73816831-73816831 on Build GRCh38
rs954271957 Chr.2: 73816694-73816694 on Build GRCh38
rs555403425 Chr.2: 73816840-73816840 on Build GRCh38
rs750487933 Chr.2: 73816659-73816659 on Build GRCh38
rs748180312 Chr.2: 73816682-73816682 on Build GRCh38
rs758443703 Chr.2: 73816852-73816852 on Build GRCh38
rs375273518 Chr.2: 73816661-73816661 on Build GRCh38
rs182355775 Chr.2: 73816829-73816829 on Build GRCh38
rs914323334 Chr.2: 73816744-73816744 on Build GRCh38
rs774729156 Chr.2: 73816817-73816817 on Build GRCh38
rs946908285 Chr.2: 73816786-73816786 on Build GRCh38
rs190141035 Chr.2: 73816669-73816669 on Build GRCh38
rs369824472 Chr.2: 73816697-73816697 on Build GRCh38
rs766535033 Chr.2: 73816833-73816833 on Build GRCh38
rs745480372 Chr.2: 73816738-73816738 on Build GRCh38
rs764517168 Chr.2: 73816802-73816802 on Build GRCh38
rs563673924 Chr.2: 73816783-73816783 on Build GRCh38
rs539780072 Chr.2: 73816730-73816730 on Build GRCh38
rs762115755 Chr.2: 73816823-73816823 on Build GRCh38
rs985622803 Chr.2: 73816722-73816722 on Build GRCh38
rs200514281 Chr.2: 73816787-73816787 on Build GRCh38
rs779964737 Chr.2: 73816677-73816677 on Build GRCh38
rs757475199 Chr.2: 73816656-73816656 on Build GRCh38
rs986657130 Chr.2: 73816673-73816673 on Build GRCh38
rs758357092 Chr.2: 73816696-73816696 on Build GRCh38
rs746765206 Chr.2: 73816705-73816705 on Build GRCh38
rs769415227 Chr.2: 73816771-73816771 on Build GRCh38
rs770733135 Chr.2: 73816726-73816726 on Build GRCh38

View Full Product Details