Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 108157063-108157290 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL4A6
Entrez Gene ID
1288
Gene Name
collagen type IV alpha 6 chain
Gene Aliases
CXDELq22.3, DELXq22.3, DFNX6
UniGene
Hs.145586
Gene Chromosome Location
Chr.X: 108155607-108439497 on Build GRCh38
Transcripts
NM_001287758.1 NM_001287759.1 NM_001287760.1 NM_001847.3 NM_033641.3 XM_006724617.3 XM_011530852.2 XM_011530853.2

SNP Detail

SNP ID Chromosome Location
rs759799172 Chr.X: 108157181-108157181 on Build GRCh38
rs774819718 Chr.X: 108157242-108157242 on Build GRCh38
rs200948393 Chr.X: 108157152-108157152 on Build GRCh38
rs767479071 Chr.X: 108157232-108157232 on Build GRCh38
rs773604789 Chr.X: 108157158-108157158 on Build GRCh38
rs776799928 Chr.X: 108157173-108157173 on Build GRCh38
rs374763127 Chr.X: 108157192-108157192 on Build GRCh38
rs868419322 Chr.X: 108157083-108157083 on Build GRCh38
rs761614268 Chr.X: 108157226-108157226 on Build GRCh38
rs573679137 Chr.X: 108157092-108157092 on Build GRCh38
rs753422300 Chr.X: 108157244-108157244 on Build GRCh38
rs766433395 Chr.X: 108157162-108157162 on Build GRCh38
rs146123040 Chr.X: 108157124-108157124 on Build GRCh38
rs765464545 Chr.X: 108157191-108157191 on Build GRCh38
rs748991025 Chr.X: 108157154-108157154 on Build GRCh38
rs748553741 Chr.X: 108157157-108157157 on Build GRCh38
rs769120116 Chr.X: 108157155-108157155 on Build GRCh38
rs756009266 Chr.X: 108157251-108157251 on Build GRCh38
rs779481851 Chr.X: 108157243-108157243 on Build GRCh38
rs777477962 Chr.X: 108157154-108157157 on Build GRCh38
rs1023483172 Chr.X: 108157237-108157237 on Build GRCh38
rs752465145 Chr.X: 108157139-108157139 on Build GRCh38
rs751229643 Chr.X: 108157266-108157269 on Build GRCh38
rs1010718048 Chr.X: 108157191-108157192 on Build GRCh38
rs780999356 Chr.X: 108157099-108157099 on Build GRCh38
rs760794179 Chr.X: 108157161-108157161 on Build GRCh38
rs757972984 Chr.X: 108157152-108157157 on Build GRCh38

View Full Product Details