Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 149621692-149621927 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF37
Entrez Gene ID
389337
Gene Name
Rho guanine nucleotide exchange factor 37
Gene Aliases
-
UniGene
Hs.256206
Gene Chromosome Location
Chr.5: 149552049-149634968 on Build GRCh38
Transcripts
NM_001001669.2 XM_005268447.4 XM_005268448.1 XM_006714784.3 XM_011537642.2 XM_017009460.1 XM_017009461.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1435093 c.1199C>T p.S400L Chr.5: 149621926-149621926 on Build GRCh38
4663170 c.1013G>A p.R338Q Chr.5: 149621740-149621740 on Build GRCh38
3612361 c.1119G>A p.R373R Chr.5: 149621846-149621846 on Build GRCh38
6604305 c.1123G>T p.E375* Chr.5: 149621850-149621850 on Build GRCh38
5620305 c.1012C>T p.R338W Chr.5: 149621739-149621739 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs560081847 Chr.5: 149621759-149621759 on Build GRCh38
rs371296533 Chr.5: 149621756-149621756 on Build GRCh38
rs769396372 Chr.5: 149621760-149621760 on Build GRCh38
rs757167558 Chr.5: 149621734-149621734 on Build GRCh38
rs774919814 Chr.5: 149621794-149621794 on Build GRCh38
rs751603664 Chr.5: 149621718-149621718 on Build GRCh38
rs778823889 Chr.5: 149621739-149621739 on Build GRCh38
rs370844005 Chr.5: 149621715-149621715 on Build GRCh38
rs78820823 Chr.5: 149621720-149621720 on Build GRCh38
rs552298903 Chr.5: 149621817-149621817 on Build GRCh38
rs762702498 Chr.5: 149621907-149621907 on Build GRCh38
rs759594287 Chr.5: 149621803-149621803 on Build GRCh38
rs865931085 Chr.5: 149621886-149621886 on Build GRCh38
rs376205167 Chr.5: 149621714-149621714 on Build GRCh38
rs760057349 Chr.5: 149621890-149621890 on Build GRCh38
rs374376221 Chr.5: 149621790-149621790 on Build GRCh38
rs758147850 Chr.5: 149621841-149621841 on Build GRCh38
rs754074265 Chr.5: 149621902-149621902 on Build GRCh38
rs923388142 Chr.5: 149621775-149621775 on Build GRCh38
rs774770878 Chr.5: 149621889-149621889 on Build GRCh38
rs771315101 Chr.5: 149621887-149621887 on Build GRCh38
rs376923718 Chr.5: 149621843-149621843 on Build GRCh38
rs750202119 Chr.5: 149621824-149621824 on Build GRCh38
rs201114208 Chr.5: 149621845-149621845 on Build GRCh38
rs747608319 Chr.5: 149621859-149621859 on Build GRCh38
rs372944428 Chr.5: 149621906-149621906 on Build GRCh38
rs752955017 Chr.5: 149621728-149621728 on Build GRCh38
rs149683598 Chr.5: 149621898-149621898 on Build GRCh38
rs769449519 Chr.5: 149621861-149621861 on Build GRCh38
rs931965994 Chr.5: 149621815-149621815 on Build GRCh38
rs746564506 Chr.5: 149621749-149621749 on Build GRCh38
rs772671320 Chr.5: 149621897-149621897 on Build GRCh38
rs139526604 Chr.5: 149621740-149621740 on Build GRCh38
rs773587777 Chr.5: 149621766-149621766 on Build GRCh38
rs200402751 Chr.5: 149621901-149621901 on Build GRCh38
rs749680482 Chr.5: 149621882-149621882 on Build GRCh38
rs537949715 Chr.5: 149621849-149621849 on Build GRCh38
rs947381063 Chr.5: 149621895-149621895 on Build GRCh38
rs775274020 Chr.5: 149621712-149621712 on Build GRCh38
rs780707855 Chr.5: 149621858-149621858 on Build GRCh38
rs758292615 Chr.5: 149621745-149621745 on Build GRCh38
rs763318423 Chr.5: 149621772-149621772 on Build GRCh38
rs779986731 Chr.5: 149621748-149621748 on Build GRCh38
rs954193300 Chr.5: 149621713-149621713 on Build GRCh38
rs369500713 Chr.5: 149621844-149621844 on Build GRCh38
rs367882590 Chr.5: 149621808-149621808 on Build GRCh38
rs748101973 Chr.5: 149621838-149621838 on Build GRCh38
rs372019080 Chr.5: 149621818-149621818 on Build GRCh38

View Full Product Details