Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 43037637-43037886 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CUL7
Entrez Gene ID
9820
Gene Name
cullin 7
Gene Aliases
3M1, KIAA0076, dJ20C7.5
UniGene
Hs.520136
Gene Chromosome Location
Chr.6: 43037617-43053950 on Build GRCh38
Transcripts
NM_001168370.1 NM_014780.4 XM_005249503.2 XM_006715285.1 XM_011515019.2 XM_011515020.2 XM_011515021.1 XM_017011533.1 XM_017011534.1 XM_017011535.1 XM_017011536.1 XM_017011537.1 XM_017011538.1 XM_017011539.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6297588 c.5050C>A p.P1684T Chr.6: 43037735-43037735 on Build GRCh38
6638172 c.4939G>A p.A1647T Chr.6: 43037846-43037846 on Build GRCh38
7423467 c.4955T>A p.V1652D Chr.6: 43037830-43037830 on Build GRCh38
4155231 c.5001C>T p.P1667P Chr.6: 43037784-43037784 on Build GRCh38
4909258 c.5253C>T p.P1751P Chr.6: 43037784-43037784 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759689995 Chr.6: 43037790-43037790 on Build GRCh38
rs773657593 Chr.6: 43037866-43037866 on Build GRCh38
rs35893653 Chr.6: 43037746-43037747 on Build GRCh38
rs779751448 Chr.6: 43037854-43037854 on Build GRCh38
rs751414053 Chr.6: 43037821-43037821 on Build GRCh38
rs758209391 Chr.6: 43037700-43037700 on Build GRCh38
rs780899726 Chr.6: 43037834-43037834 on Build GRCh38
rs116910528 Chr.6: 43037850-43037850 on Build GRCh38
rs775022798 Chr.6: 43037756-43037756 on Build GRCh38
rs73733789 Chr.6: 43037697-43037697 on Build GRCh38
rs561242234 Chr.6: 43037784-43037784 on Build GRCh38
rs780553404 Chr.6: 43037749-43037749 on Build GRCh38
rs143717016 Chr.6: 43037813-43037813 on Build GRCh38
rs201267503 Chr.6: 43037780-43037780 on Build GRCh38
rs763871170 Chr.6: 43037819-43037819 on Build GRCh38
rs150979209 Chr.6: 43037705-43037705 on Build GRCh38
rs765032085 Chr.6: 43037811-43037811 on Build GRCh38
rs936411925 Chr.6: 43037685-43037685 on Build GRCh38
rs766370002 Chr.6: 43037669-43037669 on Build GRCh38
rs769005187 Chr.6: 43037751-43037751 on Build GRCh38
rs776511309 Chr.6: 43037789-43037789 on Build GRCh38
rs745394794 Chr.6: 43037750-43037750 on Build GRCh38
rs748715099 Chr.6: 43037774-43037774 on Build GRCh38
rs758367190 Chr.6: 43037816-43037816 on Build GRCh38
rs748661881 Chr.6: 43037859-43037859 on Build GRCh38
rs561467388 Chr.6: 43037665-43037665 on Build GRCh38
rs369627224 Chr.6: 43037830-43037830 on Build GRCh38
rs773784166 Chr.6: 43037777-43037777 on Build GRCh38
rs772534400 Chr.6: 43037776-43037776 on Build GRCh38
rs556058600 Chr.6: 43037862-43037862 on Build GRCh38
rs746561312 Chr.6: 43037687-43037687 on Build GRCh38
rs754010820 Chr.6: 43037671-43037671 on Build GRCh38
rs140092102 Chr.6: 43037713-43037713 on Build GRCh38
rs754816725 Chr.6: 43037675-43037675 on Build GRCh38
rs541161018 Chr.6: 43037692-43037692 on Build GRCh38
rs772769284 Chr.6: 43037865-43037865 on Build GRCh38
rs374097275 Chr.6: 43037867-43037867 on Build GRCh38
rs375165020 Chr.6: 43037744-43037744 on Build GRCh38
rs899221507 Chr.6: 43037828-43037828 on Build GRCh38
rs202213462 Chr.6: 43037848-43037848 on Build GRCh38

View Full Product Details