Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41977878-41978107 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP1A3
Entrez Gene ID
478
Gene Name
ATPase Na+/K+ transporting subunit alpha 3
Gene Aliases
AHC2, ATP1A1, CAPOS, DYT12, RDP
UniGene
Hs.515427
Gene Chromosome Location
Chr.19: 41966582-41994276 on Build GRCh38
Transcripts
NM_001256213.1 NM_001256214.1 NM_152296.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4533146 c.1911G>A p.R637R Chr.19: 41977968-41977968 on Build GRCh38
4532837 c.1885G>A p.E629K Chr.19: 41977994-41977994 on Build GRCh38
3389055 c.1838C>T p.T613M Chr.19: 41978041-41978041 on Build GRCh38
4664278 c.1942C>T p.R648W Chr.19: 41977937-41977937 on Build GRCh38
3534553 c.1902C>T p.I634I Chr.19: 41977977-41977977 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782108634 Chr.19: 41977926-41977926 on Build GRCh38
rs782149339 Chr.19: 41978076-41978076 on Build GRCh38
rs201391210 Chr.19: 41977955-41977955 on Build GRCh38
rs782288263 Chr.19: 41978055-41978055 on Build GRCh38
rs782731051 Chr.19: 41977927-41977927 on Build GRCh38
rs376852509 Chr.19: 41978040-41978040 on Build GRCh38
rs781949719 Chr.19: 41978046-41978046 on Build GRCh38
rs782436662 Chr.19: 41977964-41977964 on Build GRCh38
rs782026217 Chr.19: 41977899-41977899 on Build GRCh38
rs782514564 Chr.19: 41977976-41977976 on Build GRCh38
rs201197776 Chr.19: 41977924-41977924 on Build GRCh38
rs781822752 Chr.19: 41977974-41977974 on Build GRCh38
rs782598926 Chr.19: 41977998-41977998 on Build GRCh38
rs782744167 Chr.19: 41977973-41977973 on Build GRCh38
rs782002906 Chr.19: 41977938-41977938 on Build GRCh38
rs782182377 Chr.19: 41978010-41978010 on Build GRCh38
rs551405286 Chr.19: 41977915-41977915 on Build GRCh38
rs782331440 Chr.19: 41977910-41977910 on Build GRCh38
rs782278209 Chr.19: 41977902-41977902 on Build GRCh38
rs144130986 Chr.19: 41977995-41977995 on Build GRCh38
rs199984360 Chr.19: 41978004-41978004 on Build GRCh38
rs80356534 Chr.19: 41978041-41978041 on Build GRCh38
rs782284444 Chr.19: 41977994-41977994 on Build GRCh38
rs782618751 Chr.19: 41977989-41977989 on Build GRCh38

View Full Product Details