Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 171689997-171690224 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYNC1I2
Entrez Gene ID
1781
Gene Name
dynein cytoplasmic 1 intermediate chain 2
Gene Aliases
DIC74, DNCI2, IC2
UniGene
Hs.546250
Gene Chromosome Location
Chr.2: 171687409-171750158 on Build GRCh38
Transcripts
NM_001271785.1 NM_001271786.1 NM_001271787.1 NM_001271788.1 NM_001271789.1 NM_001271790.1 NM_001320882.1 NM_001320883.1 NM_001320884.1 NM_001378.2 XM_006712347.3 XM_017003525.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4086754 c.54G>T p.L18L Chr.2: 171690209-171690209 on Build GRCh38
1009546 c.38G>A p.R13H Chr.2: 171690193-171690193 on Build GRCh38
1237090 c.22A>C p.K8Q Chr.2: 171690177-171690177 on Build GRCh38
4086755 c.54G>T p.L18L Chr.2: 171690209-171690209 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs974990166 Chr.2: 171690105-171690105 on Build GRCh38
rs540814246 Chr.2: 171690204-171690204 on Build GRCh38
rs1018901471 Chr.2: 171690061-171690061 on Build GRCh38
rs368799638 Chr.2: 171690136-171690136 on Build GRCh38
rs562892372 Chr.2: 171690154-171690154 on Build GRCh38
rs866506642 Chr.2: 171690180-171690180 on Build GRCh38
rs756352854 Chr.2: 171690108-171690108 on Build GRCh38
rs1023443082 Chr.2: 171690093-171690093 on Build GRCh38
rs185747503 Chr.2: 171690125-171690125 on Build GRCh38
rs780419839 Chr.2: 171690125-171690127 on Build GRCh38
rs922178901 Chr.2: 171690134-171690134 on Build GRCh38

View Full Product Details