Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 69082772-69083019 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA6
Entrez Gene ID
23460
Gene Name
ATP binding cassette subfamily A member 6
Gene Aliases
EST155051
UniGene
Hs.709514
Gene Chromosome Location
Chr.17: 69062044-69142188 on Build GRCh38
Transcripts
NM_080284.2 XM_011524574.2 XM_017024404.1 XM_017024405.1 XM_017024406.1 XM_017024407.1 XM_017024408.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3521403 c.4476-1G>A p.? Chr.17: 69083014-69083014 on Build GRCh38
5956380 c.4519G>A p.D1507N Chr.17: 69082970-69082970 on Build GRCh38
3403158 c.4506C>G p.N1502K Chr.17: 69082983-69082983 on Build GRCh38
3521402 c.4560G>A p.V1520V Chr.17: 69082929-69082929 on Build GRCh38
707475 c.4595C>T p.P1532L Chr.17: 69082894-69082894 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777337889 Chr.17: 69082837-69082837 on Build GRCh38
rs972155702 Chr.17: 69082861-69082861 on Build GRCh38
rs765199719 Chr.17: 69082887-69082887 on Build GRCh38
rs892673825 Chr.17: 69082971-69082971 on Build GRCh38
rs759377509 Chr.17: 69082934-69082934 on Build GRCh38
rs753405366 Chr.17: 69082948-69082948 on Build GRCh38
rs184811201 Chr.17: 69082886-69082886 on Build GRCh38
rs762003028 Chr.17: 69082885-69082885 on Build GRCh38
rs779518531 Chr.17: 69082852-69082852 on Build GRCh38
rs751580826 Chr.17: 69082902-69082902 on Build GRCh38
rs745329177 Chr.17: 69082984-69082984 on Build GRCh38
rs769032135 Chr.17: 69082873-69082873 on Build GRCh38
rs370155388 Chr.17: 69082863-69082863 on Build GRCh38
rs776944594 Chr.17: 69082881-69082881 on Build GRCh38
rs755775187 Chr.17: 69082832-69082832 on Build GRCh38
rs868185042 Chr.17: 69082919-69082919 on Build GRCh38
rs779990705 Chr.17: 69082970-69082970 on Build GRCh38
rs776012885 Chr.17: 69082866-69082866 on Build GRCh38
rs758415619 Chr.17: 69082919-69082919 on Build GRCh38
rs772626326 Chr.17: 69082865-69082865 on Build GRCh38
rs78383857 Chr.17: 69082938-69082938 on Build GRCh38
rs752402814 Chr.17: 69082828-69082828 on Build GRCh38
rs941654510 Chr.17: 69082995-69082995 on Build GRCh38
rs905161771 Chr.17: 69082931-69082931 on Build GRCh38
rs748858676 Chr.17: 69082839-69082839 on Build GRCh38
rs377673526 Chr.17: 69082939-69082939 on Build GRCh38
rs867902458 Chr.17: 69082896-69082896 on Build GRCh38
rs1011549109 Chr.17: 69082804-69082804 on Build GRCh38
rs746361809 Chr.17: 69082855-69082855 on Build GRCh38
rs767382213 Chr.17: 69082936-69082936 on Build GRCh38
rs909477769 Chr.17: 69082976-69082976 on Build GRCh38
rs374780972 Chr.17: 69082842-69082842 on Build GRCh38
rs778277038 Chr.17: 69082967-69082967 on Build GRCh38

View Full Product Details