Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 151743621-151743860 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNGA2
Entrez Gene ID
1260
Gene Name
cyclic nucleotide gated channel alpha 2
Gene Aliases
CNCA, CNCA1, CNG2, OCNC1, OCNCALPHA, OCNCa
UniGene
Hs.447360
Gene Chromosome Location
Chr.X: 151734746-151745564 on Build GRCh38
Transcripts
NM_005140.1

SNP Detail

SNP ID Chromosome Location
rs201839589 Chr.X: 151743657-151743657 on Build GRCh38
rs770232273 Chr.X: 151743800-151743800 on Build GRCh38
rs747110357 Chr.X: 151743764-151743764 on Build GRCh38
rs776787921 Chr.X: 151743793-151743793 on Build GRCh38
rs763518565 Chr.X: 151743811-151743811 on Build GRCh38
rs372801718 Chr.X: 151743693-151743693 on Build GRCh38
rs747214127 Chr.X: 151743656-151743656 on Build GRCh38
rs200939277 Chr.X: 151743767-151743767 on Build GRCh38
rs764557841 Chr.X: 151743822-151743822 on Build GRCh38
rs199869471 Chr.X: 151743796-151743796 on Build GRCh38
rs147982375 Chr.X: 151743648-151743648 on Build GRCh38
rs371987829 Chr.X: 151743677-151743677 on Build GRCh38
rs200521778 Chr.X: 151743683-151743683 on Build GRCh38
rs375775509 Chr.X: 151743832-151743832 on Build GRCh38
rs150539917 Chr.X: 151743699-151743699 on Build GRCh38
rs375373217 Chr.X: 151743671-151743671 on Build GRCh38
rs777882225 Chr.X: 151743747-151743747 on Build GRCh38
rs772082842 Chr.X: 151743647-151743647 on Build GRCh38
rs754939798 Chr.X: 151743722-151743722 on Build GRCh38
rs776025165 Chr.X: 151743805-151743805 on Build GRCh38
rs950021351 Chr.X: 151743667-151743667 on Build GRCh38
rs746099774 Chr.X: 151743799-151743799 on Build GRCh38
rs763616745 Chr.X: 151743686-151743686 on Build GRCh38
rs749175350 Chr.X: 151743721-151743721 on Build GRCh38
rs141614710 Chr.X: 151743698-151743698 on Build GRCh38
rs755841985 Chr.X: 151743708-151743708 on Build GRCh38
rs866573072 Chr.X: 151743739-151743739 on Build GRCh38
rs867991029 Chr.X: 151743763-151743763 on Build GRCh38
rs866130959 Chr.X: 151743719-151743719 on Build GRCh38
rs765837704 Chr.X: 151743675-151743675 on Build GRCh38
rs771102269 Chr.X: 151743670-151743670 on Build GRCh38
rs924411492 Chr.X: 151743672-151743672 on Build GRCh38
rs773897889 Chr.X: 151743831-151743831 on Build GRCh38
rs866580601 Chr.X: 151743782-151743782 on Build GRCh38
rs779838845 Chr.X: 151743715-151743715 on Build GRCh38
rs367749104 Chr.X: 151743834-151743834 on Build GRCh38
rs751067612 Chr.X: 151743688-151743688 on Build GRCh38

View Full Product Details