Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 99997506-99997735 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTS17
Entrez Gene ID
170691
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 17
Gene Aliases
-
UniGene
Hs.513200
Gene Chromosome Location
Chr.15: 99971438-100342924 on Build GRCh38
Transcripts
NM_139057.3 XM_005254872.3 XM_011521312.2 XM_017021973.1 XM_017021974.1 XM_017021975.1 XM_017021976.1 XM_017021978.1 XM_017021979.1 XM_017021980.1 XM_017021982.1 XM_017021983.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1740201 c.2650C>T p.R884W Chr.15: 99997531-99997531 on Build GRCh38
6594006 c.2606C>T p.P869L Chr.15: 99997575-99997575 on Build GRCh38
6594014 c.2621C>T p.S874L Chr.15: 99997560-99997560 on Build GRCh38
5473595 c.2651G>A p.R884Q Chr.15: 99997530-99997530 on Build GRCh38
6245451 c.2606C>A p.P869Q Chr.15: 99997575-99997575 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs563938783 Chr.15: 99997544-99997544 on Build GRCh38
rs1008859063 Chr.15: 99997693-99997693 on Build GRCh38
rs779698939 Chr.15: 99997610-99997610 on Build GRCh38
rs751012843 Chr.15: 99997599-99997599 on Build GRCh38
rs776124364 Chr.15: 99997597-99997597 on Build GRCh38
rs375519462 Chr.15: 99997608-99997608 on Build GRCh38
rs773881815 Chr.15: 99997633-99997633 on Build GRCh38
rs569576215 Chr.15: 99997669-99997669 on Build GRCh38
rs760236403 Chr.15: 99997639-99997639 on Build GRCh38
rs982823860 Chr.15: 99997666-99997666 on Build GRCh38
rs113426361 Chr.15: 99997661-99997661 on Build GRCh38
rs780077915 Chr.15: 99997556-99997556 on Build GRCh38
rs922631314 Chr.15: 99997530-99997530 on Build GRCh38
rs977439848 Chr.15: 99997660-99997660 on Build GRCh38
rs1055650935 Chr.15: 99997589-99997589 on Build GRCh38
rs750044969 Chr.15: 99997607-99997607 on Build GRCh38
rs747049463 Chr.15: 99997620-99997620 on Build GRCh38
rs780979498 Chr.15: 99997541-99997541 on Build GRCh38
rs140577767 Chr.15: 99997583-99997583 on Build GRCh38
rs779560660 Chr.15: 99997662-99997662 on Build GRCh38
rs917184116 Chr.15: 99997644-99997644 on Build GRCh38
rs533467085 Chr.15: 99997698-99997698 on Build GRCh38
rs957131238 Chr.15: 99997641-99997641 on Build GRCh38
rs775650700 Chr.15: 99997624-99997624 on Build GRCh38
rs749156998 Chr.15: 99997557-99997557 on Build GRCh38
rs768473488 Chr.15: 99997607-99997610 on Build GRCh38
rs550989841 Chr.15: 99997656-99997656 on Build GRCh38
rs754802167 Chr.15: 99997616-99997616 on Build GRCh38
rs756837804 Chr.15: 99997531-99997531 on Build GRCh38
rs746067605 Chr.15: 99997623-99997623 on Build GRCh38
rs143323522 Chr.15: 99997547-99997547 on Build GRCh38
rs765499241 Chr.15: 99997596-99997596 on Build GRCh38
rs778009417 Chr.15: 99997619-99997619 on Build GRCh38
rs112856607 Chr.15: 99997645-99997645 on Build GRCh38
rs377387249 Chr.15: 99997560-99997560 on Build GRCh38
rs369460266 Chr.15: 99997622-99997622 on Build GRCh38
rs773418364 Chr.15: 99997559-99997559 on Build GRCh38
rs755357010 Chr.15: 99997654-99997654 on Build GRCh38
rs151305163 Chr.15: 99997574-99997574 on Build GRCh38
rs572716880 Chr.15: 99997554-99997554 on Build GRCh38
rs766989655 Chr.15: 99997637-99997637 on Build GRCh38
rs769102110 Chr.15: 99997632-99997632 on Build GRCh38
rs952237627 Chr.15: 99997614-99997614 on Build GRCh38
rs760021311 Chr.15: 99997575-99997575 on Build GRCh38
rs753876970 Chr.15: 99997615-99997615 on Build GRCh38
rs202079882 Chr.15: 99997605-99997605 on Build GRCh38
rs761476488 Chr.15: 99997601-99997601 on Build GRCh38
rs373353462 Chr.15: 99997598-99997598 on Build GRCh38
rs891850525 Chr.15: 99997681-99997681 on Build GRCh38

View Full Product Details