Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 115127576-115127827 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATRNL1
Entrez Gene ID
26033
Gene Name
attractin like 1
Gene Aliases
ALP, bA338L11.1, bA454H24.1
UniGene
Hs.501127
Gene Chromosome Location
Chr.10: 115093365-115948999 on Build GRCh38
Transcripts
NM_001276282.3 NM_207303.4 NR_074088.2 XM_011539587.1 XM_011539588.2 XM_011539589.2 XM_011539590.2 XM_011539591.2 XM_017016035.1 XM_017016036.1 XM_017016037.1 XM_017016038.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
915093 c.518G>A p.G173D Chr.10: 115127619-115127619 on Build GRCh38
683224 c.554G>A p.G185D Chr.10: 115127655-115127655 on Build GRCh38
6128485 c.522T>A p.N174K Chr.10: 115127623-115127623 on Build GRCh38
3434417 c.505C>T p.P169S Chr.10: 115127606-115127606 on Build GRCh38
2054365 c.568C>A p.H190N Chr.10: 115127669-115127669 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781860851 Chr.10: 115127622-115127622 on Build GRCh38
rs139960943 Chr.10: 115127689-115127689 on Build GRCh38
rs782086664 Chr.10: 115127742-115127742 on Build GRCh38
rs369461649 Chr.10: 115127739-115127739 on Build GRCh38
rs782691406 Chr.10: 115127734-115127734 on Build GRCh38
rs782072853 Chr.10: 115127732-115127732 on Build GRCh38
rs782162083 Chr.10: 115127729-115127729 on Build GRCh38
rs782272283 Chr.10: 115127669-115127669 on Build GRCh38
rs782620826 Chr.10: 115127649-115127649 on Build GRCh38
rs111592092 Chr.10: 115127733-115127733 on Build GRCh38
rs782502499 Chr.10: 115127656-115127656 on Build GRCh38
rs782549906 Chr.10: 115127642-115127642 on Build GRCh38
rs781900461 Chr.10: 115127770-115127770 on Build GRCh38
rs781996026 Chr.10: 115127703-115127703 on Build GRCh38
rs543682519 Chr.10: 115127735-115127735 on Build GRCh38
rs781959120 Chr.10: 115127618-115127618 on Build GRCh38
rs782775721 Chr.10: 115127619-115127619 on Build GRCh38
rs782337434 Chr.10: 115127700-115127700 on Build GRCh38
rs782670374 Chr.10: 115127666-115127666 on Build GRCh38
rs782781233 Chr.10: 115127764-115127764 on Build GRCh38
rs781814517 Chr.10: 115127643-115127643 on Build GRCh38
rs782439386 Chr.10: 115127648-115127648 on Build GRCh38
rs368755603 Chr.10: 115127721-115127721 on Build GRCh38
rs782792235 Chr.10: 115127734-115127735 on Build GRCh38
rs782217624 Chr.10: 115127654-115127654 on Build GRCh38
rs782223880 Chr.10: 115127698-115127698 on Build GRCh38
rs372430190 Chr.10: 115127722-115127722 on Build GRCh38
rs782693801 Chr.10: 115127616-115127616 on Build GRCh38
rs782122735 Chr.10: 115127719-115127719 on Build GRCh38

View Full Product Details