Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 55727236-55727467 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD63
Entrez Gene ID
967
Gene Name
CD63 molecule
Gene Aliases
LAMP-3, ME491, MLA1, OMA81H, TSPAN30
UniGene
Hs.445570
Gene Chromosome Location
Chr.12: 55725443-55729673 on Build GRCh38
Transcripts
NM_001257389.1 NM_001257390.1 NM_001257391.1 NM_001257392.1 NM_001257400.1 NM_001257401.1 NM_001267698.1 NM_001780.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3463260 c.139G>A p.A47T Chr.12: 55727267-55727267 on Build GRCh38
6218105 c.136G>T p.G46W Chr.12: 55727270-55727270 on Build GRCh38
3463261 c.71G>A p.C24Y Chr.12: 55727335-55727335 on Build GRCh38
4622691 c.110T>G p.L37R Chr.12: 55727296-55727296 on Build GRCh38
468638 c.126C>T p.T42T Chr.12: 55727280-55727280 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs576536793 Chr.12: 55727390-55727390 on Build GRCh38
rs922843447 Chr.12: 55727264-55727264 on Build GRCh38
rs574316609 Chr.12: 55727400-55727400 on Build GRCh38
rs771912345 Chr.12: 55727317-55727317 on Build GRCh38
rs548153645 Chr.12: 55727262-55727262 on Build GRCh38
rs775885379 Chr.12: 55727349-55727349 on Build GRCh38
rs982762976 Chr.12: 55727339-55727339 on Build GRCh38
rs768363181 Chr.12: 55727304-55727304 on Build GRCh38
rs757540937 Chr.12: 55727259-55727259 on Build GRCh38
rs977334983 Chr.12: 55727261-55727261 on Build GRCh38
rs878967218 Chr.12: 55727340-55727340 on Build GRCh38
rs569909052 Chr.12: 55727297-55727297 on Build GRCh38
rs755977795 Chr.12: 55727271-55727271 on Build GRCh38
rs765784784 Chr.12: 55727344-55727344 on Build GRCh38
rs150015050 Chr.12: 55727315-55727315 on Build GRCh38
rs919347497 Chr.12: 55727431-55727431 on Build GRCh38
rs749059145 Chr.12: 55727303-55727303 on Build GRCh38
rs959951003 Chr.12: 55727402-55727402 on Build GRCh38
rs112219729 Chr.12: 55727299-55727299 on Build GRCh38
rs772257 Chr.12: 55727410-55727410 on Build GRCh38
rs1051723644 Chr.12: 55727441-55727441 on Build GRCh38
rs764328479 Chr.12: 55727256-55727256 on Build GRCh38
rs751761251 Chr.12: 55727257-55727257 on Build GRCh38
rs760270529 Chr.12: 55727322-55727322 on Build GRCh38
rs941372277 Chr.12: 55727376-55727376 on Build GRCh38
rs772847145 Chr.12: 55727319-55727319 on Build GRCh38
rs113863438 Chr.12: 55727359-55727359 on Build GRCh38
rs534368854 Chr.12: 55727397-55727397 on Build GRCh38
rs991395005 Chr.12: 55727407-55727407 on Build GRCh38
rs963524853 Chr.12: 55727411-55727411 on Build GRCh38
rs778749476 Chr.12: 55727295-55727296 on Build GRCh38
rs749396682 Chr.12: 55727259-55727259 on Build GRCh38
rs1021782091 Chr.12: 55727307-55727307 on Build GRCh38
rs145161311 Chr.12: 55727399-55727399 on Build GRCh38
rs752005937 Chr.12: 55727385-55727385 on Build GRCh38
rs764336878 Chr.12: 55727370-55727370 on Build GRCh38
rs905277127 Chr.12: 55727396-55727396 on Build GRCh38
rs144910349 Chr.12: 55727267-55727267 on Build GRCh38
rs774239613 Chr.12: 55727312-55727312 on Build GRCh38
rs878991886 Chr.12: 55727341-55727341 on Build GRCh38
rs929369126 Chr.12: 55727434-55727434 on Build GRCh38
rs973541479 Chr.12: 55727420-55727420 on Build GRCh38
rs767729381 Chr.12: 55727391-55727391 on Build GRCh38

View Full Product Details