Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 3080834-3081077 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARSF
Entrez Gene ID
416
Gene Name
arylsulfatase F
Gene Aliases
ASF
UniGene
Hs.101674
Gene Chromosome Location
Chr.X: 3040234-3112729 on Build GRCh38
Transcripts
NM_001201538.1 NM_001201539.1 NM_004042.4 XM_011545522.1 XM_011545523.1 XM_011545524.1 XM_017029527.1 XM_017029528.1

SNP Detail

SNP ID Chromosome Location
rs750328635 Chr.X: 3080901-3080901 on Build GRCh38
rs764101471 Chr.X: 3081014-3081014 on Build GRCh38
rs768393004 Chr.X: 3080979-3080979 on Build GRCh38
rs143530347 Chr.X: 3080940-3080940 on Build GRCh38
rs772718607 Chr.X: 3080957-3080957 on Build GRCh38
rs188839908 Chr.X: 3080934-3080934 on Build GRCh38
rs889662886 Chr.X: 3080910-3080910 on Build GRCh38
rs757425032 Chr.X: 3081039-3081039 on Build GRCh38
rs775143829 Chr.X: 3080867-3080867 on Build GRCh38
rs772891680 Chr.X: 3081003-3081003 on Build GRCh38
rs776396879 Chr.X: 3080982-3080982 on Build GRCh38
rs201567455 Chr.X: 3081006-3081006 on Build GRCh38
rs765580113 Chr.X: 3081051-3081051 on Build GRCh38
rs199739800 Chr.X: 3080951-3080951 on Build GRCh38
rs751005503 Chr.X: 3080878-3080878 on Build GRCh38
rs768761561 Chr.X: 3080930-3080930 on Build GRCh38
rs375489043 Chr.X: 3080893-3080893 on Build GRCh38
rs746585324 Chr.X: 3080965-3080965 on Build GRCh38
rs965998855 Chr.X: 3080945-3080945 on Build GRCh38
rs747045341 Chr.X: 3080928-3080928 on Build GRCh38
rs867577161 Chr.X: 3080914-3080914 on Build GRCh38
rs367880082 Chr.X: 3081011-3081011 on Build GRCh38
rs200244823 Chr.X: 3080865-3080865 on Build GRCh38
rs183383325 Chr.X: 3080920-3080920 on Build GRCh38
rs772377819 Chr.X: 3080969-3080969 on Build GRCh38
rs368462218 Chr.X: 3080896-3080896 on Build GRCh38
rs754127728 Chr.X: 3080939-3080939 on Build GRCh38
rs371667500 Chr.X: 3081020-3081020 on Build GRCh38
rs767522990 Chr.X: 3080927-3080927 on Build GRCh38
rs779698098 Chr.X: 3080963-3080963 on Build GRCh38
rs768482550 Chr.X: 3080884-3080884 on Build GRCh38
rs375328193 Chr.X: 3081004-3081004 on Build GRCh38
rs779174946 Chr.X: 3080941-3080941 on Build GRCh38
rs938794959 Chr.X: 3081013-3081029 on Build GRCh38
rs573237918 Chr.X: 3080885-3080885 on Build GRCh38
rs138366402 Chr.X: 3081002-3081002 on Build GRCh38
rs747780230 Chr.X: 3080994-3080994 on Build GRCh38
rs1021026528 Chr.X: 3080961-3080961 on Build GRCh38

View Full Product Details