Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 29327197-29327460 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BACH1
Entrez Gene ID
571
Gene Name
BTB domain and CNC homolog 1
Gene Aliases
BACH-1, BTBD24
UniGene
Hs.154276
Gene Chromosome Location
Chr.21: 29298795-29361896 on Build GRCh38
Transcripts
NM_001186.3 NM_206866.2 NR_027655.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6161062 c.1417T>A p.S473T Chr.21: 29327241-29327241 on Build GRCh38
4435851 c.1442A>T p.D481V Chr.21: 29327266-29327266 on Build GRCh38
297792 c.1523C>T p.T508I Chr.21: 29327347-29327347 on Build GRCh38
1737754 c.1543T>C p.C515R Chr.21: 29327367-29327367 on Build GRCh38
6190165 c.1561G>C p.E521Q Chr.21: 29327385-29327385 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761553877 Chr.21: 29327242-29327242 on Build GRCh38
rs780743906 Chr.21: 29327283-29327283 on Build GRCh38
rs762912319 Chr.21: 29327258-29327258 on Build GRCh38
rs768892267 Chr.21: 29327335-29327335 on Build GRCh38
rs375374696 Chr.21: 29327376-29327376 on Build GRCh38
rs754615744 Chr.21: 29327274-29327274 on Build GRCh38
rs575620663 Chr.21: 29327230-29327230 on Build GRCh38
rs754204780 Chr.21: 29327296-29327296 on Build GRCh38
rs779363849 Chr.21: 29327302-29327302 on Build GRCh38
rs911049235 Chr.21: 29327299-29327299 on Build GRCh38
rs746309353 Chr.21: 29327303-29327303 on Build GRCh38
rs762823839 Chr.21: 29327360-29327360 on Build GRCh38
rs751087323 Chr.21: 29327265-29327265 on Build GRCh38
rs142524709 Chr.21: 29327264-29327264 on Build GRCh38
rs902242107 Chr.21: 29327261-29327261 on Build GRCh38
rs755794107 Chr.21: 29327404-29327404 on Build GRCh38
rs544651776 Chr.21: 29327375-29327375 on Build GRCh38
rs199802427 Chr.21: 29327244-29327244 on Build GRCh38
rs779086628 Chr.21: 29327411-29327411 on Build GRCh38
rs140585235 Chr.21: 29327336-29327336 on Build GRCh38
rs780267098 Chr.21: 29327317-29327317 on Build GRCh38
rs367693467 Chr.21: 29327358-29327358 on Build GRCh38
rs751034308 Chr.21: 29327371-29327371 on Build GRCh38
rs371330530 Chr.21: 29327366-29327366 on Build GRCh38
rs530151866 Chr.21: 29327247-29327247 on Build GRCh38
rs752248312 Chr.21: 29327382-29327382 on Build GRCh38
rs148401992 Chr.21: 29327239-29327239 on Build GRCh38
rs143227772 Chr.21: 29327227-29327227 on Build GRCh38
rs201560153 Chr.21: 29327330-29327330 on Build GRCh38
rs35766001 Chr.21: 29327236-29327236 on Build GRCh38
rs750875639 Chr.21: 29327415-29327415 on Build GRCh38
rs1049633413 Chr.21: 29327267-29327267 on Build GRCh38
rs762119140 Chr.21: 29327342-29327342 on Build GRCh38
rs151321858 Chr.21: 29327316-29327316 on Build GRCh38
rs757513028 Chr.21: 29327301-29327301 on Build GRCh38
rs769438465 Chr.21: 29327348-29327348 on Build GRCh38
rs775714503 Chr.21: 29327235-29327235 on Build GRCh38
rs769056682 Chr.21: 29327238-29327238 on Build GRCh38

View Full Product Details