Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 112109434-112109707 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3orf52
Entrez Gene ID
79669
Gene Name
chromosome 3 open reading frame 52
Gene Aliases
TTMP
UniGene
Hs.434247
Gene Chromosome Location
Chr.3: 112086335-112131095 on Build GRCh38
Transcripts
NM_001171747.1 NM_024616.2 XM_011513133.2 XM_017007181.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4112219 c.424T>G p.S142A Chr.3: 112109570-112109570 on Build GRCh38
5689530 c.452A>G p.E151G Chr.3: 112109598-112109598 on Build GRCh38
4968995 c.406G>C p.V136L Chr.3: 112109552-112109552 on Build GRCh38
5447524 c.440T>C p.F147S Chr.3: 112109586-112109586 on Build GRCh38
3721075 c.399C>A p.L133L Chr.3: 112109545-112109545 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs869301439 Chr.3: 112109538-112109539 on Build GRCh38
rs971913455 Chr.3: 112109625-112109625 on Build GRCh38
rs758958652 Chr.3: 112109510-112109510 on Build GRCh38
rs754247906 Chr.3: 112109500-112109500 on Build GRCh38
rs775144134 Chr.3: 112109585-112109589 on Build GRCh38
rs972102977 Chr.3: 112109466-112109466 on Build GRCh38
rs375590614 Chr.3: 112109575-112109575 on Build GRCh38
rs2120095 Chr.3: 112109629-112109629 on Build GRCh38
rs966268121 Chr.3: 112109584-112109584 on Build GRCh38
rs869213992 Chr.3: 112109541-112109542 on Build GRCh38
rs778671591 Chr.3: 112109639-112109639 on Build GRCh38
rs779867317 Chr.3: 112109620-112109620 on Build GRCh38
rs340167 Chr.3: 112109576-112109576 on Build GRCh38
rs78063105 Chr.3: 112109676-112109676 on Build GRCh38
rs765843751 Chr.3: 112109505-112109505 on Build GRCh38
rs749293566 Chr.3: 112109537-112109537 on Build GRCh38
rs780914069 Chr.3: 112109597-112109597 on Build GRCh38
rs919342223 Chr.3: 112109482-112109482 on Build GRCh38
rs775231258 Chr.3: 112109514-112109514 on Build GRCh38
rs370777296 Chr.3: 112109594-112109594 on Build GRCh38
rs960983413 Chr.3: 112109654-112109654 on Build GRCh38
rs771653423 Chr.3: 112109557-112109558 on Build GRCh38
rs142875002 Chr.3: 112109555-112109555 on Build GRCh38
rs5851818 Chr.3: 112109537-112109538 on Build GRCh38
rs1000917132 Chr.3: 112109496-112109496 on Build GRCh38
rs768546198 Chr.3: 112109546-112109546 on Build GRCh38
rs766651353 Chr.3: 112109566-112109566 on Build GRCh38
rs904872742 Chr.3: 112109590-112109590 on Build GRCh38
rs775254538 Chr.3: 112109533-112109533 on Build GRCh38
rs745680611 Chr.3: 112109527-112109527 on Build GRCh38
rs542099104 Chr.3: 112109475-112109475 on Build GRCh38
rs397949663 Chr.3: 112109540-112109541 on Build GRCh38
rs903974275 Chr.3: 112109487-112109487 on Build GRCh38
rs761711287 Chr.3: 112109565-112109565 on Build GRCh38
rs563854279 Chr.3: 112109569-112109569 on Build GRCh38
rs368126508 Chr.3: 112109580-112109580 on Build GRCh38
rs755351434 Chr.3: 112109504-112109504 on Build GRCh38
rs753197993 Chr.3: 112109506-112109506 on Build GRCh38
rs200500372 Chr.3: 112109512-112109512 on Build GRCh38
rs769525120 Chr.3: 112109530-112109530 on Build GRCh38
rs565822581 Chr.3: 112109641-112109641 on Build GRCh38
rs139638398 Chr.3: 112109667-112109668 on Build GRCh38
rs368290990 Chr.3: 112109659-112109659 on Build GRCh38
rs770857041 Chr.3: 112109653-112109653 on Build GRCh38
rs148943408 Chr.3: 112109517-112109517 on Build GRCh38
rs746480575 Chr.3: 112109513-112109513 on Build GRCh38
rs559883651 Chr.3: 112109606-112109606 on Build GRCh38
rs952140175 Chr.3: 112109680-112109680 on Build GRCh38
rs373918440 Chr.3: 112109579-112109579 on Build GRCh38
rs749057542 Chr.3: 112109628-112109628 on Build GRCh38
rs182004669 Chr.3: 112109619-112109619 on Build GRCh38
rs373442818 Chr.3: 112109570-112109570 on Build GRCh38

View Full Product Details