Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7454186-7454419 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNB1
Entrez Gene ID
1140
Gene Name
cholinergic receptor nicotinic beta 1 subunit
Gene Aliases
ACHRB, CHRNB, CMS1D, CMS2A, CMS2C, SCCMS
UniGene
Hs.330386
Gene Chromosome Location
Chr.17: 7445087-7457613 on Build GRCh38
Transcripts
NM_000747.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
984391 c.852G>A p.L284L Chr.17: 7454328-7454328 on Build GRCh38
6082048 c.838T>C p.S280P Chr.17: 7454314-7454314 on Build GRCh38
6991563 c.864T>C p.T288T Chr.17: 7454340-7454340 on Build GRCh38
5572564 c.931T>G p.Y311D Chr.17: 7454407-7454407 on Build GRCh38
4510796 c.849C>T p.A283A Chr.17: 7454325-7454325 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148217194 Chr.17: 7454383-7454383 on Build GRCh38
rs773696133 Chr.17: 7454256-7454256 on Build GRCh38
rs748428224 Chr.17: 7454371-7454371 on Build GRCh38
rs781475180 Chr.17: 7454364-7454364 on Build GRCh38
rs763393615 Chr.17: 7454258-7454258 on Build GRCh38
rs753695361 Chr.17: 7454316-7454316 on Build GRCh38
rs757194127 Chr.17: 7454317-7454317 on Build GRCh38
rs778746961 Chr.17: 7454318-7454318 on Build GRCh38
rs756960602 Chr.17: 7454355-7454358 on Build GRCh38
rs373928636 Chr.17: 7454246-7454246 on Build GRCh38
rs137852811 Chr.17: 7454329-7454329 on Build GRCh38
rs755060667 Chr.17: 7454350-7454350 on Build GRCh38
rs769813899 Chr.17: 7454273-7454273 on Build GRCh38
rs188543727 Chr.17: 7454238-7454238 on Build GRCh38
rs764035317 Chr.17: 7454293-7454293 on Build GRCh38
rs371190837 Chr.17: 7454281-7454281 on Build GRCh38
rs758993114 Chr.17: 7454271-7454271 on Build GRCh38
rs945086307 Chr.17: 7454374-7454374 on Build GRCh38
rs956543933 Chr.17: 7454353-7454353 on Build GRCh38
rs137852810 Chr.17: 7454341-7454341 on Build GRCh38
rs998494409 Chr.17: 7454319-7454320 on Build GRCh38
rs117168441 Chr.17: 7454379-7454379 on Build GRCh38
rs140169734 Chr.17: 7454377-7454377 on Build GRCh38
rs961734949 Chr.17: 7454230-7454230 on Build GRCh38
rs780391043 Chr.17: 7454358-7454358 on Build GRCh38
rs201129045 Chr.17: 7454323-7454323 on Build GRCh38
rs753428978 Chr.17: 7454329-7454334 on Build GRCh38
rs989527059 Chr.17: 7454326-7454326 on Build GRCh38
rs112895222 Chr.17: 7454277-7454277 on Build GRCh38
rs770017785 Chr.17: 7454375-7454375 on Build GRCh38
rs567864992 Chr.17: 7454266-7454266 on Build GRCh38
rs1024444772 Chr.17: 7454252-7454252 on Build GRCh38
rs774773039 Chr.17: 7454385-7454385 on Build GRCh38
rs180908813 Chr.17: 7454255-7454255 on Build GRCh38
rs145826933 Chr.17: 7454328-7454328 on Build GRCh38
rs202089107 Chr.17: 7454262-7454262 on Build GRCh38
rs76001008 Chr.17: 7454288-7454288 on Build GRCh38

View Full Product Details