Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 27401126-27401399 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARNTL2
Entrez Gene ID
56938
Gene Name
aryl hydrocarbon receptor nuclear translocator like 2
Gene Aliases
BMAL2, CLIF, MOP9, PASD9, bHLHe6
UniGene
Hs.445447
Gene Chromosome Location
Chr.12: 27332854-27425813 on Build GRCh38
Transcripts
NM_001248002.1 NM_001248003.1 NM_001248004.1 NM_001248005.1 NM_020183.4 XM_006719112.3 XM_006719114.3 XM_011520766.2 XM_011520768.2 XM_011520769.2 XM_011520770.2 XM_011520771.2 XM_017019666.1 XM_017019667.1 XM_017019668.1 XM_017019669.1 NR_109975.1

Gene Symbol
ARNTL2-AS1
Entrez Gene ID
101928646
Gene Name
ARNTL2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.434269
Gene Chromosome Location
Chr.12: 27389789-27446634 on Build GRCh38
Transcripts
NM_001248002.1 NM_001248003.1 NM_001248004.1 NM_001248005.1 NM_020183.4 XM_006719112.3 XM_006719114.3 XM_011520766.2 XM_011520768.2 XM_011520769.2 XM_011520770.2 XM_011520771.2 XM_017019666.1 XM_017019667.1 XM_017019668.1 XM_017019669.1 NR_109975.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7325860 c.1192G>T p.E398* Chr.12: 27401288-27401288 on Build GRCh38
5761483 c.1213T>G p.Y405D Chr.12: 27401309-27401309 on Build GRCh38
3459817 c.1178G>A p.G393E Chr.12: 27401274-27401274 on Build GRCh38
938399 c.1190A>G p.Q397R Chr.12: 27401286-27401286 on Build GRCh38
3687877 c.1264G>A p.A422T Chr.12: 27401360-27401360 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs151042635 Chr.12: 27401150-27401150 on Build GRCh38
rs570623981 Chr.12: 27401170-27401170 on Build GRCh38
rs964651721 Chr.12: 27401265-27401265 on Build GRCh38
rs757994593 Chr.12: 27401260-27401260 on Build GRCh38
rs757518739 Chr.12: 27401291-27401291 on Build GRCh38
rs770405883 Chr.12: 27401329-27401329 on Build GRCh38
rs749310622 Chr.12: 27401372-27401372 on Build GRCh38
rs753735092 Chr.12: 27401227-27401227 on Build GRCh38
rs532927468 Chr.12: 27401258-27401258 on Build GRCh38
rs747383259 Chr.12: 27401200-27401200 on Build GRCh38
rs561373413 Chr.12: 27401165-27401165 on Build GRCh38
rs895293312 Chr.12: 27401159-27401159 on Build GRCh38
rs114189648 Chr.12: 27401207-27401207 on Build GRCh38
rs562452045 Chr.12: 27401244-27401244 on Build GRCh38
rs760326010 Chr.12: 27401216-27401216 on Build GRCh38
rs190015013 Chr.12: 27401163-27401163 on Build GRCh38
rs939478290 Chr.12: 27401173-27401173 on Build GRCh38
rs889047269 Chr.12: 27401354-27401354 on Build GRCh38
rs780771594 Chr.12: 27401363-27401363 on Build GRCh38
rs377480542 Chr.12: 27401310-27401310 on Build GRCh38
rs774633807 Chr.12: 27401214-27401214 on Build GRCh38
rs757354805 Chr.12: 27401172-27401172 on Build GRCh38
rs1032310713 Chr.12: 27401189-27401189 on Build GRCh38
rs765268417 Chr.12: 27401328-27401329 on Build GRCh38
rs1033905290 Chr.12: 27401153-27401153 on Build GRCh38
rs759437442 Chr.12: 27401236-27401236 on Build GRCh38
rs762070443 Chr.12: 27401216-27401222 on Build GRCh38
rs751238912 Chr.12: 27401279-27401279 on Build GRCh38
rs201450723 Chr.12: 27401224-27401224 on Build GRCh38
rs777225804 Chr.12: 27401266-27401266 on Build GRCh38
rs923137005 Chr.12: 27401325-27401325 on Build GRCh38
rs781716894 Chr.12: 27401305-27401305 on Build GRCh38
rs990044137 Chr.12: 27401257-27401257 on Build GRCh38

View Full Product Details