Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 7737382-7737643 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC4C
Entrez Gene ID
170482
Gene Name
C-type lectin domain family 4 member C
Gene Aliases
BDCA2, CD303, CLECSF11, CLECSF7, DLEC, HECL, PRO34150
UniGene
Hs.351812
Gene Chromosome Location
Chr.12: 7729245-7749473 on Build GRCh38
Transcripts
NM_130441.2 NM_203503.1 XM_017018940.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6880585 c.330C>T p.N110N Chr.12: 7737480-7737480 on Build GRCh38
1639258 c.250C>A p.P84T Chr.12: 7737560-7737560 on Build GRCh38
1705094 c.260G>A p.W87* Chr.12: 7737550-7737550 on Build GRCh38
7152922 c.236-1G>T p.? Chr.12: 7737575-7737575 on Build GRCh38
5970850 c.278G>A p.S93N Chr.12: 7737532-7737532 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749952691 Chr.12: 7737464-7737464 on Build GRCh38
rs762505849 Chr.12: 7737591-7737591 on Build GRCh38
rs760446580 Chr.12: 7737420-7737420 on Build GRCh38
rs781712344 Chr.12: 7737474-7737474 on Build GRCh38
rs932183665 Chr.12: 7737429-7737429 on Build GRCh38
rs925255697 Chr.12: 7737568-7737568 on Build GRCh38
rs763901195 Chr.12: 7737575-7737575 on Build GRCh38
rs748670892 Chr.12: 7737476-7737476 on Build GRCh38
rs77889141 Chr.12: 7737444-7737444 on Build GRCh38
rs750068065 Chr.12: 7737590-7737590 on Build GRCh38
rs756676359 Chr.12: 7737607-7737607 on Build GRCh38
rs759638243 Chr.12: 7737410-7737410 on Build GRCh38
rs774655325 Chr.12: 7737527-7737527 on Build GRCh38
rs372099582 Chr.12: 7737413-7737413 on Build GRCh38
rs747389242 Chr.12: 7737435-7737444 on Build GRCh38
rs1015000247 Chr.12: 7737428-7737428 on Build GRCh38
rs749576045 Chr.12: 7737504-7737504 on Build GRCh38
rs778167358 Chr.12: 7737486-7737486 on Build GRCh38
rs746013818 Chr.12: 7737543-7737543 on Build GRCh38
rs776516470 Chr.12: 7737577-7737577 on Build GRCh38
rs771299204 Chr.12: 7737522-7737522 on Build GRCh38
rs764877133 Chr.12: 7737583-7737583 on Build GRCh38
rs768942691 Chr.12: 7737464-7737464 on Build GRCh38
rs61747156 Chr.12: 7737480-7737480 on Build GRCh38
rs201723542 Chr.12: 7737598-7737598 on Build GRCh38
rs764823955 Chr.12: 7737457-7737457 on Build GRCh38
rs953189667 Chr.12: 7737492-7737492 on Build GRCh38
rs781714231 Chr.12: 7737529-7737530 on Build GRCh38
rs760629901 Chr.12: 7737566-7737566 on Build GRCh38
rs763926863 Chr.12: 7737440-7737440 on Build GRCh38
rs772267983 Chr.12: 7737556-7737556 on Build GRCh38
rs143057533 Chr.12: 7737571-7737571 on Build GRCh38
rs757833588 Chr.12: 7737465-7737465 on Build GRCh38
rs756969299 Chr.12: 7737448-7737448 on Build GRCh38
rs986468730 Chr.12: 7737555-7737555 on Build GRCh38
rs777752009 Chr.12: 7737604-7737604 on Build GRCh38
rs761540827 Chr.12: 7737578-7737578 on Build GRCh38
rs142924521 Chr.12: 7737554-7737554 on Build GRCh38

View Full Product Details