Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 96115260-96115509 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADRA2B
Entrez Gene ID
151
Gene Name
adrenoceptor alpha 2B
Gene Aliases
ADRA2L1, ADRA2RL1, ADRARL1, ALPHA2BAR, FAME2, alpha-2BAR
UniGene
Hs.247686
Gene Chromosome Location
Chr.2: 96112875-96116245 on Build GRCh38
Transcripts
NM_000682.6

SNP Detail

SNP ID Chromosome Location
rs753633590 Chr.2: 96115298-96115298 on Build GRCh38
rs772737564 Chr.2: 96115396-96115396 on Build GRCh38
rs780332816 Chr.2: 96115353-96115353 on Build GRCh38
rs756639557 Chr.2: 96115464-96115464 on Build GRCh38
rs201255251 Chr.2: 96115348-96115348 on Build GRCh38
rs770303610 Chr.2: 96115352-96115352 on Build GRCh38
rs766876470 Chr.2: 96115290-96115297 on Build GRCh38
rs760565402 Chr.2: 96115401-96115401 on Build GRCh38
rs771504647 Chr.2: 96115393-96115393 on Build GRCh38
rs36097995 Chr.2: 96115478-96115478 on Build GRCh38
rs776725267 Chr.2: 96115490-96115490 on Build GRCh38
rs915691407 Chr.2: 96115338-96115338 on Build GRCh38
rs553348015 Chr.2: 96115314-96115314 on Build GRCh38
rs781173591 Chr.2: 96115462-96115462 on Build GRCh38
rs770838322 Chr.2: 96115489-96115489 on Build GRCh38
rs761395837 Chr.2: 96115388-96115388 on Build GRCh38
rs752212429 Chr.2: 96115382-96115384 on Build GRCh38
rs779198872 Chr.2: 96115480-96115480 on Build GRCh38
rs768420641 Chr.2: 96115485-96115485 on Build GRCh38
rs751767748 Chr.2: 96115321-96115321 on Build GRCh38
rs556445952 Chr.2: 96115443-96115443 on Build GRCh38
rs1041338935 Chr.2: 96115410-96115410 on Build GRCh38
rs766114681 Chr.2: 96115283-96115283 on Build GRCh38
rs1049900125 Chr.2: 96115455-96115455 on Build GRCh38
rs766314237 Chr.2: 96115419-96115419 on Build GRCh38
rs786205528 Chr.2: 96115486-96115486 on Build GRCh38
rs759399024 Chr.2: 96115434-96115434 on Build GRCh38
rs776448757 Chr.2: 96115492-96115492 on Build GRCh38
rs758739163 Chr.2: 96115308-96115308 on Build GRCh38
rs781287713 Chr.2: 96115346-96115346 on Build GRCh38
rs756029699 Chr.2: 96115281-96115281 on Build GRCh38
rs931061984 Chr.2: 96115339-96115339 on Build GRCh38
rs780609027 Chr.2: 96115473-96115473 on Build GRCh38
rs879255577 Chr.2: 96115464-96115475 on Build GRCh38
rs757428840 Chr.2: 96115345-96115345 on Build GRCh38
rs765069734 Chr.2: 96115436-96115436 on Build GRCh38
rs747828766 Chr.2: 96115482-96115482 on Build GRCh38
rs750494938 Chr.2: 96115463-96115463 on Build GRCh38
rs753630708 Chr.2: 96115420-96115420 on Build GRCh38
rs959938122 Chr.2: 96115295-96115295 on Build GRCh38
rs545761782 Chr.2: 96115363-96115363 on Build GRCh38
rs768365911 Chr.2: 96115373-96115373 on Build GRCh38
rs773995734 Chr.2: 96115376-96115376 on Build GRCh38
rs866930463 Chr.2: 96115347-96115347 on Build GRCh38
rs928447398 Chr.2: 96115292-96115292 on Build GRCh38
rs368576695 Chr.2: 96115452-96115452 on Build GRCh38

View Full Product Details