Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 47357825-47358060 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHX34
Entrez Gene ID
9704
Gene Name
DEAH-box helicase 34
Gene Aliases
DDX34, HRH1
UniGene
Hs.151706
Gene Chromosome Location
Chr.19: 47349281-47382704 on Build GRCh38
Transcripts
NM_014681.5 XM_005259443.3 XM_011527550.2 XM_011527551.2 XM_017027524.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3362931 c.1082C>T p.P361L Chr.19: 47357930-47357930 on Build GRCh38
7189211 c.1185G>T p.L395L Chr.19: 47358033-47358033 on Build GRCh38
3536320 c.1133G>T p.R378L Chr.19: 47357981-47357981 on Build GRCh38
6643540 c.1204G>A p.A402T Chr.19: 47358052-47358052 on Build GRCh38
3536319 c.1110C>T p.D370D Chr.19: 47357958-47357958 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201521695 Chr.19: 47357900-47357900 on Build GRCh38
rs764158084 Chr.19: 47357849-47357849 on Build GRCh38
rs757302035 Chr.19: 47357969-47357969 on Build GRCh38
rs757965971 Chr.19: 47357994-47357994 on Build GRCh38
rs969938719 Chr.19: 47357899-47357899 on Build GRCh38
rs778289113 Chr.19: 47357916-47357916 on Build GRCh38
rs760640285 Chr.19: 47357848-47357848 on Build GRCh38
rs766416052 Chr.19: 47357941-47357941 on Build GRCh38
rs369619890 Chr.19: 47357927-47357927 on Build GRCh38
rs150672057 Chr.19: 47357925-47357925 on Build GRCh38
rs754172613 Chr.19: 47358031-47358031 on Build GRCh38
rs768568236 Chr.19: 47358000-47358000 on Build GRCh38
rs748061566 Chr.19: 47358011-47358011 on Build GRCh38
rs375719793 Chr.19: 47357988-47357988 on Build GRCh38
rs778434752 Chr.19: 47357980-47357980 on Build GRCh38
rs761725648 Chr.19: 47357940-47357940 on Build GRCh38
rs372827547 Chr.19: 47357954-47357954 on Build GRCh38
rs774308379 Chr.19: 47357952-47357952 on Build GRCh38
rs774454068 Chr.19: 47358015-47358015 on Build GRCh38
rs147969937 Chr.19: 47358028-47358028 on Build GRCh38
rs144919221 Chr.19: 47357995-47357995 on Build GRCh38
rs774574790 Chr.19: 47357910-47357911 on Build GRCh38
rs753812770 Chr.19: 47357959-47357959 on Build GRCh38
rs759758867 Chr.19: 47357953-47357953 on Build GRCh38
rs751803740 Chr.19: 47357874-47357874 on Build GRCh38
rs775436190 Chr.19: 47357844-47357844 on Build GRCh38
rs200869108 Chr.19: 47358006-47358006 on Build GRCh38
rs756577133 Chr.19: 47357894-47357894 on Build GRCh38
rs756452016 Chr.19: 47357967-47357967 on Build GRCh38
rs200822007 Chr.19: 47357963-47357963 on Build GRCh38
rs750986492 Chr.19: 47358035-47358035 on Build GRCh38
rs770467047 Chr.19: 47357907-47357907 on Build GRCh38
rs759508384 Chr.19: 47358018-47358018 on Build GRCh38
rs369014615 Chr.19: 47357901-47357901 on Build GRCh38
rs765630323 Chr.19: 47358033-47358033 on Build GRCh38
rs115851317 Chr.19: 47357970-47357970 on Build GRCh38
rs113135692 Chr.19: 47357888-47357888 on Build GRCh38
rs767635576 Chr.19: 47358024-47358024 on Build GRCh38
rs746767339 Chr.19: 47357926-47357926 on Build GRCh38
rs907222293 Chr.19: 47357922-47357922 on Build GRCh38
rs866949481 Chr.19: 47357987-47357987 on Build GRCh38
rs903106077 Chr.19: 47357981-47357981 on Build GRCh38
rs142904187 Chr.19: 47357889-47357889 on Build GRCh38
rs776698450 Chr.19: 47357933-47357933 on Build GRCh38
rs757739030 Chr.19: 47358032-47358032 on Build GRCh38
rs745463102 Chr.19: 47357924-47357924 on Build GRCh38
rs746094820 Chr.19: 47358014-47358014 on Build GRCh38
rs972393251 Chr.19: 47358025-47358025 on Build GRCh38
rs763101376 Chr.19: 47357861-47357861 on Build GRCh38
rs891742850 Chr.19: 47357935-47357935 on Build GRCh38
rs760888791 Chr.19: 47358027-47358027 on Build GRCh38
rs371822195 Chr.19: 47357879-47357879 on Build GRCh38
rs755924998 Chr.19: 47357931-47357931 on Build GRCh38
rs201682143 Chr.19: 47357854-47357854 on Build GRCh38
rs766427726 Chr.19: 47357867-47357867 on Build GRCh38
rs962075067 Chr.19: 47357903-47357903 on Build GRCh38
rs1049268889 Chr.19: 47357880-47357880 on Build GRCh38

View Full Product Details