Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 19362300-19362561 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B9D1
Entrez Gene ID
27077
Gene Name
B9 domain containing 1
Gene Aliases
B9, EPPB9, MKS9, MKSR1
UniGene
Hs.309467
Gene Chromosome Location
Chr.17: 19335372-19378193 on Build GRCh38
Transcripts
NM_001243473.2 NM_001243475.2 NM_001321214.1 NM_001321215.1 NM_001321216.1 NM_001321217.1 NM_001321218.1 NM_001321219.1 NM_015681.4 XM_005256607.2 XM_005256610.1 XM_017024452.1

SNP Detail

SNP ID Chromosome Location
rs770918238 Chr.17: 19362472-19362472 on Build GRCh38
rs976412640 Chr.17: 19362430-19362430 on Build GRCh38
rs1046425051 Chr.17: 19362542-19362542 on Build GRCh38
rs746112614 Chr.17: 19362476-19362476 on Build GRCh38
rs1014924936 Chr.17: 19362356-19362356 on Build GRCh38
rs774736264 Chr.17: 19362480-19362480 on Build GRCh38
rs573443036 Chr.17: 19362345-19362345 on Build GRCh38
rs10445411 Chr.17: 19362484-19362484 on Build GRCh38
rs770185431 Chr.17: 19362477-19362477 on Build GRCh38
rs1011941761 Chr.17: 19362431-19362431 on Build GRCh38
rs1045371925 Chr.17: 19362399-19362399 on Build GRCh38
rs751480715 Chr.17: 19362537-19362537 on Build GRCh38
rs922599692 Chr.17: 19362443-19362443 on Build GRCh38
rs758139304 Chr.17: 19362521-19362521 on Build GRCh38
rs759167462 Chr.17: 19362493-19362493 on Build GRCh38
rs1004705386 Chr.17: 19362419-19362419 on Build GRCh38
rs954051262 Chr.17: 19362460-19362460 on Build GRCh38
rs985529436 Chr.17: 19362463-19362463 on Build GRCh38
rs751608872 Chr.17: 19362501-19362501 on Build GRCh38
rs1015252200 Chr.17: 19362420-19362420 on Build GRCh38
rs764022966 Chr.17: 19362529-19362529 on Build GRCh38
rs781395275 Chr.17: 19362540-19362540 on Build GRCh38
rs747379585 Chr.17: 19362448-19362448 on Build GRCh38
rs1030639955 Chr.17: 19362478-19362478 on Build GRCh38
rs765907063 Chr.17: 19362487-19362487 on Build GRCh38
rs775969961 Chr.17: 19362478-19362478 on Build GRCh38
rs752474351 Chr.17: 19362514-19362514 on Build GRCh38
rs769196411 Chr.17: 19362479-19362479 on Build GRCh38
rs748772003 Chr.17: 19362461-19362470 on Build GRCh38
rs776218520 Chr.17: 19362488-19362488 on Build GRCh38
rs747052229 Chr.17: 19362465-19362465 on Build GRCh38
rs140375234 Chr.17: 19362415-19362415 on Build GRCh38
rs764936291 Chr.17: 19362500-19362500 on Build GRCh38
rs541848827 Chr.17: 19362400-19362400 on Build GRCh38
rs1001214347 Chr.17: 19362414-19362414 on Build GRCh38
rs1044830946 Chr.17: 19362448-19362449 on Build GRCh38
rs781521965 Chr.17: 19362475-19362475 on Build GRCh38
rs965949224 Chr.17: 19362421-19362421 on Build GRCh38
rs951866292 Chr.17: 19362417-19362417 on Build GRCh38

View Full Product Details