Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 85873230-85873463 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC83
Entrez Gene ID
220047
Gene Name
coiled-coil domain containing 83
Gene Aliases
CT148, HSD9
UniGene
Hs.567774
Gene Chromosome Location
Chr.11: 85855066-85920021 on Build GRCh38
Transcripts
NM_001286159.1 NM_173556.4 XM_011544839.2 XM_011544840.2 XM_011544841.1 XM_011544842.1 XM_011544843.1 XM_017017354.1 XM_017017355.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6328063 c.117C>T p.A39A Chr.11: 85873232-85873232 on Build GRCh38
6519421 c.124C>A p.Q42K Chr.11: 85873239-85873239 on Build GRCh38
1628268 c.135T>A p.F45L Chr.11: 85873250-85873250 on Build GRCh38
6519422 c.132G>A p.M44I Chr.11: 85873247-85873247 on Build GRCh38
2221503 c.152G>T p.R51M Chr.11: 85873267-85873267 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs12362209 Chr.11: 85873260-85873260 on Build GRCh38
rs754574178 Chr.11: 85873327-85873327 on Build GRCh38
rs367629145 Chr.11: 85873319-85873319 on Build GRCh38
rs992289175 Chr.11: 85873302-85873302 on Build GRCh38
rs768772015 Chr.11: 85873317-85873318 on Build GRCh38
rs767449764 Chr.11: 85873320-85873320 on Build GRCh38
rs747320696 Chr.11: 85873288-85873288 on Build GRCh38
rs755803121 Chr.11: 85873323-85873323 on Build GRCh38
rs748542491 Chr.11: 85873325-85873326 on Build GRCh38
rs768878602 Chr.11: 85873316-85873316 on Build GRCh38
rs746922200 Chr.11: 85873363-85873363 on Build GRCh38
rs750102971 Chr.11: 85873251-85873251 on Build GRCh38
rs143421054 Chr.11: 85873309-85873309 on Build GRCh38
rs1033326527 Chr.11: 85873271-85873271 on Build GRCh38
rs751574927 Chr.11: 85873317-85873317 on Build GRCh38
rs1041141597 Chr.11: 85873258-85873258 on Build GRCh38
rs771297572 Chr.11: 85873289-85873289 on Build GRCh38
rs879174681 Chr.11: 85873396-85873396 on Build GRCh38
rs778082744 Chr.11: 85873341-85873341 on Build GRCh38
rs200852045 Chr.11: 85873315-85873315 on Build GRCh38
rs1006114373 Chr.11: 85873358-85873358 on Build GRCh38
rs772426367 Chr.11: 85873276-85873276 on Build GRCh38
rs759786985 Chr.11: 85873303-85873303 on Build GRCh38
rs781467986 Chr.11: 85873323-85873324 on Build GRCh38
rs143615039 Chr.11: 85873277-85873277 on Build GRCh38
rs925238475 Chr.11: 85873383-85873383 on Build GRCh38
rs750043930 Chr.11: 85873322-85873322 on Build GRCh38
rs775411167 Chr.11: 85873312-85873312 on Build GRCh38
rs779732713 Chr.11: 85873269-85873269 on Build GRCh38
rs763137267 Chr.11: 85873314-85873314 on Build GRCh38
rs868597305 Chr.11: 85873412-85873412 on Build GRCh38
rs936581325 Chr.11: 85873385-85873385 on Build GRCh38
rs905706447 Chr.11: 85873388-85873388 on Build GRCh38
rs753282698 Chr.11: 85873326-85873326 on Build GRCh38
rs776769203 Chr.11: 85873299-85873299 on Build GRCh38
rs1035524204 Chr.11: 85873425-85873425 on Build GRCh38
rs1044677799 Chr.11: 85873371-85873371 on Build GRCh38
rs527549546 Chr.11: 85873274-85873274 on Build GRCh38

View Full Product Details