Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 123019126-123019385 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DERL1
Entrez Gene ID
79139
Gene Name
derlin 1
Gene Aliases
DER-1, DER1
UniGene
Hs.241576
Gene Chromosome Location
Chr.8: 123013164-123042423 on Build GRCh38
Transcripts
NM_001134671.2 NM_024295.5 XM_006716657.1 XM_011517299.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1095681 c.519G>T p.E173D Chr.8: 123019293-123019293 on Build GRCh38
1095680 c.562T>G p.F188V Chr.8: 123019250-123019250 on Build GRCh38
4781573 c.547T>A p.Y183N Chr.8: 123019265-123019265 on Build GRCh38
5661720 c.610C>T p.Q204* Chr.8: 123019202-123019202 on Build GRCh38
3644749 c.555C>T p.F185F Chr.8: 123019257-123019257 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs201953852 Chr.8: 123019346-123019346 on Build GRCh38
rs561331450 Chr.8: 123019201-123019201 on Build GRCh38
rs770586414 Chr.8: 123019252-123019252 on Build GRCh38
rs776418046 Chr.8: 123019170-123019170 on Build GRCh38
rs372216708 Chr.8: 123019172-123019172 on Build GRCh38
rs372267337 Chr.8: 123019238-123019238 on Build GRCh38
rs894103549 Chr.8: 123019224-123019224 on Build GRCh38
rs751887904 Chr.8: 123019207-123019207 on Build GRCh38
rs767779165 Chr.8: 123019213-123019213 on Build GRCh38
rs377249453 Chr.8: 123019308-123019308 on Build GRCh38
rs2272722 Chr.8: 123019301-123019301 on Build GRCh38
rs757547996 Chr.8: 123019212-123019212 on Build GRCh38
rs139154983 Chr.8: 123019204-123019204 on Build GRCh38
rs375483266 Chr.8: 123019192-123019192 on Build GRCh38
rs200911646 Chr.8: 123019214-123019214 on Build GRCh38
rs772901228 Chr.8: 123019161-123019161 on Build GRCh38
rs550573125 Chr.8: 123019317-123019317 on Build GRCh38
rs201119908 Chr.8: 123019293-123019293 on Build GRCh38
rs771451866 Chr.8: 123019157-123019157 on Build GRCh38
rs765239113 Chr.8: 123019180-123019180 on Build GRCh38
rs571672566 Chr.8: 123019232-123019232 on Build GRCh38
rs200467011 Chr.8: 123019268-123019268 on Build GRCh38
rs746655677 Chr.8: 123019165-123019165 on Build GRCh38
rs546867515 Chr.8: 123019309-123019309 on Build GRCh38
rs752313640 Chr.8: 123019326-123019327 on Build GRCh38
rs764884711 Chr.8: 123019312-123019313 on Build GRCh38
rs761924697 Chr.8: 123019337-123019337 on Build GRCh38
rs770782961 Chr.8: 123019169-123019169 on Build GRCh38
rs550182260 Chr.8: 123019243-123019243 on Build GRCh38
rs200376396 Chr.8: 123019297-123019297 on Build GRCh38
rs780652342 Chr.8: 123019229-123019229 on Build GRCh38

View Full Product Details