Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 47503149-47503412 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD2AP
Entrez Gene ID
23607
Gene Name
CD2 associated protein
Gene Aliases
CMS
UniGene
Hs.485518
Gene Chromosome Location
Chr.6: 47477746-47627263 on Build GRCh38
Transcripts
NM_012120.2 XM_005248976.1 XM_017010641.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3874582 c.86T>C p.V29A Chr.6: 47503361-47503361 on Build GRCh38
1319083 c.137G>T p.R46I Chr.6: 47503412-47503412 on Build GRCh38
5822713 c.5-2A>C p.? Chr.6: 47503278-47503278 on Build GRCh38
279716 c.62G>A p.R21Q Chr.6: 47503337-47503337 on Build GRCh38
3995031 c.104A>T p.E35V Chr.6: 47503379-47503379 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs187865496 Chr.6: 47503179-47503179 on Build GRCh38
rs760269835 Chr.6: 47503260-47503261 on Build GRCh38
rs572687654 Chr.6: 47503284-47503284 on Build GRCh38
rs772360705 Chr.6: 47503382-47503382 on Build GRCh38
rs746124377 Chr.6: 47503360-47503360 on Build GRCh38
rs559978848 Chr.6: 47503337-47503337 on Build GRCh38
rs758262794 Chr.6: 47503371-47503371 on Build GRCh38
rs367952819 Chr.6: 47503301-47503301 on Build GRCh38
rs767423641 Chr.6: 47503286-47503286 on Build GRCh38
rs751278706 Chr.6: 47503272-47503272 on Build GRCh38
rs996396457 Chr.6: 47503215-47503215 on Build GRCh38
rs752519964 Chr.6: 47503300-47503300 on Build GRCh38
rs34773184 Chr.6: 47503317-47503317 on Build GRCh38
rs777711947 Chr.6: 47503304-47503304 on Build GRCh38
rs772843395 Chr.6: 47503262-47503262 on Build GRCh38
rs746283610 Chr.6: 47503234-47503234 on Build GRCh38
rs941934411 Chr.6: 47503241-47503241 on Build GRCh38
rs773187230 Chr.6: 47503259-47503259 on Build GRCh38
rs769170284 Chr.6: 47503261-47503261 on Build GRCh38
rs900756601 Chr.6: 47503181-47503183 on Build GRCh38
rs770750324 Chr.6: 47503267-47503268 on Build GRCh38
rs774669667 Chr.6: 47503229-47503229 on Build GRCh38
rs1028005495 Chr.6: 47503228-47503228 on Build GRCh38
rs762661708 Chr.6: 47503266-47503266 on Build GRCh38
rs776778555 Chr.6: 47503303-47503304 on Build GRCh38
rs891177463 Chr.6: 47503289-47503289 on Build GRCh38
rs970462010 Chr.6: 47503171-47503171 on Build GRCh38
rs779061496 Chr.6: 47503353-47503353 on Build GRCh38
rs530231846 Chr.6: 47503270-47503270 on Build GRCh38
rs3756935 Chr.6: 47503260-47503260 on Build GRCh38
rs772451959 Chr.6: 47503245-47503245 on Build GRCh38

View Full Product Details