Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27136905-27137178 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRR30
Entrez Gene ID
339779
Gene Name
proline rich 30
Gene Aliases
C2orf53
UniGene
Hs.136555
Gene Chromosome Location
Chr.2: 27136847-27139464 on Build GRCh38
Transcripts
NM_178553.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6091021 c.1178C>A p.P393H Chr.2: 27137152-27137152 on Build GRCh38
1326248 c.1163C>A p.S388Y Chr.2: 27137167-27137167 on Build GRCh38
3910115 c.1155C>T p.V385V Chr.2: 27137175-27137175 on Build GRCh38
6247809 c.1206T>C p.S402S Chr.2: 27137124-27137124 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs535804301 Chr.2: 27136966-27136966 on Build GRCh38
rs1038995272 Chr.2: 27137095-27137095 on Build GRCh38
rs868625599 Chr.2: 27136999-27136999 on Build GRCh38
rs11121 Chr.2: 27137019-27137019 on Build GRCh38
rs779028584 Chr.2: 27137124-27137124 on Build GRCh38
rs928833920 Chr.2: 27137026-27137026 on Build GRCh38
rs774392192 Chr.2: 27137050-27137050 on Build GRCh38
rs898495445 Chr.2: 27136946-27136946 on Build GRCh38
rs745877581 Chr.2: 27137133-27137133 on Build GRCh38
rs764892131 Chr.2: 27137075-27137075 on Build GRCh38
rs1032056114 Chr.2: 27137058-27137058 on Build GRCh38
rs779714404 Chr.2: 27137106-27137106 on Build GRCh38
rs776110340 Chr.2: 27137072-27137072 on Build GRCh38
rs957306547 Chr.2: 27136942-27136942 on Build GRCh38
rs568368172 Chr.2: 27136945-27136945 on Build GRCh38
rs145730256 Chr.2: 27136983-27136983 on Build GRCh38
rs757456813 Chr.2: 27137121-27137121 on Build GRCh38
rs543693262 Chr.2: 27137079-27137079 on Build GRCh38
rs564072931 Chr.2: 27137078-27137078 on Build GRCh38
rs769101100 Chr.2: 27136936-27136938 on Build GRCh38
rs772559822 Chr.2: 27137059-27137059 on Build GRCh38
rs916158744 Chr.2: 27136984-27136984 on Build GRCh38
rs771038419 Chr.2: 27137043-27137043 on Build GRCh38
rs929455494 Chr.2: 27137028-27137028 on Build GRCh38
rs111711012 Chr.2: 27136966-27136966 on Build GRCh38
rs772645137 Chr.2: 27137144-27137144 on Build GRCh38
rs558366083 Chr.2: 27137101-27137101 on Build GRCh38
rs371665403 Chr.2: 27137100-27137100 on Build GRCh38
rs1048314556 Chr.2: 27137038-27137038 on Build GRCh38
rs1013528248 Chr.2: 27137009-27137009 on Build GRCh38
rs912713887 Chr.2: 27137138-27137138 on Build GRCh38
rs760887297 Chr.2: 27137061-27137061 on Build GRCh38
rs751384314 Chr.2: 27137093-27137093 on Build GRCh38
rs987099710 Chr.2: 27137125-27137125 on Build GRCh38
rs766165599 Chr.2: 27137083-27137083 on Build GRCh38
rs569342063 Chr.2: 27136997-27136997 on Build GRCh38
rs139568064 Chr.2: 27136940-27136940 on Build GRCh38
rs1046647785 Chr.2: 27137015-27137015 on Build GRCh38
rs755625236 Chr.2: 27137060-27137060 on Build GRCh38
rs761530162 Chr.2: 27137073-27137073 on Build GRCh38
rs763832661 Chr.2: 27137068-27137068 on Build GRCh38
rs909290841 Chr.2: 27137066-27137066 on Build GRCh38

View Full Product Details