Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 34319401-34319674 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCL8
Entrez Gene ID
6355
Gene Name
C-C motif chemokine ligand 8
Gene Aliases
HC14, MCP-2, MCP2, SCYA10, SCYA8
UniGene
Hs.271387
Gene Chromosome Location
Chr.17: 34319047-34321402 on Build GRCh38
Transcripts
NM_005623.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3180811 c.20T>G p.L7R Chr.17: 34319521-34319521 on Build GRCh38
7360079 c.41C>T p.A14V Chr.17: 34319542-34319542 on Build GRCh38
4065464 c.30G>A p.L10L Chr.17: 34319531-34319531 on Build GRCh38
5342907 c.18G>A p.A6A Chr.17: 34319519-34319519 on Build GRCh38
6080612 c.33G>T p.L11L Chr.17: 34319534-34319534 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765971174 Chr.17: 34319493-34319493 on Build GRCh38
rs115582326 Chr.17: 34319427-34319427 on Build GRCh38
rs972653173 Chr.17: 34319634-34319634 on Build GRCh38
rs775968153 Chr.17: 34319560-34319560 on Build GRCh38
rs200372834 Chr.17: 34319572-34319572 on Build GRCh38
rs779943250 Chr.17: 34319469-34319469 on Build GRCh38
rs774153890 Chr.17: 34319640-34319640 on Build GRCh38
rs768645764 Chr.17: 34319482-34319482 on Build GRCh38
rs1041611986 Chr.17: 34319508-34319508 on Build GRCh38
rs757458292 Chr.17: 34319518-34319518 on Build GRCh38
rs755839542 Chr.17: 34319464-34319464 on Build GRCh38
rs199714946 Chr.17: 34319481-34319481 on Build GRCh38
rs766235535 Chr.17: 34319508-34319509 on Build GRCh38
rs201103777 Chr.17: 34319505-34319505 on Build GRCh38
rs751514779 Chr.17: 34319578-34319579 on Build GRCh38
rs200483515 Chr.17: 34319545-34319545 on Build GRCh38
rs754816900 Chr.17: 34319619-34319619 on Build GRCh38
rs35401229 Chr.17: 34319542-34319542 on Build GRCh38
rs758522235 Chr.17: 34319454-34319454 on Build GRCh38
rs200602745 Chr.17: 34319443-34319443 on Build GRCh38
rs771469853 Chr.17: 34319533-34319533 on Build GRCh38
rs200100731 Chr.17: 34319500-34319500 on Build GRCh38
rs201631782 Chr.17: 34319490-34319490 on Build GRCh38
rs376503806 Chr.17: 34319608-34319608 on Build GRCh38
rs759868755 Chr.17: 34319555-34319555 on Build GRCh38
rs200834045 Chr.17: 34319483-34319483 on Build GRCh38
rs375015363 Chr.17: 34319487-34319487 on Build GRCh38
rs148233199 Chr.17: 34319528-34319528 on Build GRCh38
rs150734427 Chr.17: 34319557-34319557 on Build GRCh38
rs773770346 Chr.17: 34319504-34319506 on Build GRCh38
rs375474786 Chr.17: 34319599-34319599 on Build GRCh38
rs772709607 Chr.17: 34319489-34319489 on Build GRCh38
rs113350491 Chr.17: 34319450-34319450 on Build GRCh38
rs199536140 Chr.17: 34319452-34319452 on Build GRCh38
rs777278201 Chr.17: 34319541-34319541 on Build GRCh38
rs201281336 Chr.17: 34319627-34319627 on Build GRCh38
rs750716076 Chr.17: 34319617-34319617 on Build GRCh38
rs764516836 Chr.17: 34319565-34319565 on Build GRCh38
rs752224481 Chr.17: 34319639-34319639 on Build GRCh38
rs771679076 Chr.17: 34319484-34319484 on Build GRCh38
rs554461653 Chr.17: 34319552-34319552 on Build GRCh38
rs201624360 Chr.17: 34319561-34319561 on Build GRCh38
rs199868476 Chr.17: 34319428-34319428 on Build GRCh38
rs761823803 Chr.17: 34319586-34319586 on Build GRCh38
rs776943192 Chr.17: 34319495-34319495 on Build GRCh38
rs766806815 Chr.17: 34319435-34319435 on Build GRCh38
rs574575546 Chr.17: 34319585-34319585 on Build GRCh38
rs569040565 Chr.17: 34319525-34319525 on Build GRCh38
rs751609965 Chr.17: 34319515-34319515 on Build GRCh38
rs754322738 Chr.17: 34319436-34319436 on Build GRCh38
rs35694639 Chr.17: 34319454-34319455 on Build GRCh38
rs202065406 Chr.17: 34319461-34319461 on Build GRCh38
rs539862720 Chr.17: 34319498-34319498 on Build GRCh38
rs974568028 Chr.17: 34319476-34319476 on Build GRCh38
rs143666282 Chr.17: 34319519-34319519 on Build GRCh38
rs755430338 Chr.17: 34319441-34319441 on Build GRCh38
rs764279272 Chr.17: 34319514-34319514 on Build GRCh38
rs781547852 Chr.17: 34319423-34319423 on Build GRCh38
rs767832524 Chr.17: 34319591-34319591 on Build GRCh38
rs34918489 Chr.17: 34319523-34319523 on Build GRCh38
rs200651459 Chr.17: 34319459-34319459 on Build GRCh38

View Full Product Details