Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 67483114-67483385 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AIP
Entrez Gene ID
9049
Gene Name
aryl hydrocarbon receptor interacting protein
Gene Aliases
ARA9, FKBP16, FKBP37, SMTPHN, XAP-2, XAP2
UniGene
Hs.412433
Gene Chromosome Location
Chr.11: 67483029-67491108 on Build GRCh38
Transcripts
NM_001302960.1 NM_003977.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3810223 c.12C>T p.I4I Chr.11: 67483170-67483170 on Build GRCh38
5839384 c.41delA p.K15fs*3 Chr.11: 67483199-67483199 on Build GRCh38
234375 c.75C>T p.L25L Chr.11: 67483233-67483233 on Build GRCh38
7205655 c.60A>G p.E20E Chr.11: 67483218-67483218 on Build GRCh38
7148039 c.51G>A p.V17V Chr.11: 67483209-67483209 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374324200 Chr.11: 67483248-67483248 on Build GRCh38
rs200665479 Chr.11: 67483136-67483136 on Build GRCh38
rs766095172 Chr.11: 67483300-67483300 on Build GRCh38
rs886037871 Chr.11: 67483161-67483161 on Build GRCh38
rs984976766 Chr.11: 67483319-67483319 on Build GRCh38
rs779881664 Chr.11: 67483262-67483262 on Build GRCh38
rs940872517 Chr.11: 67483328-67483328 on Build GRCh38
rs267606547 Chr.11: 67483161-67483162 on Build GRCh38
rs767063262 Chr.11: 67483294-67483294 on Build GRCh38
rs776131484 Chr.11: 67483236-67483236 on Build GRCh38
rs371423932 Chr.11: 67483242-67483242 on Build GRCh38
rs761198473 Chr.11: 67483291-67483291 on Build GRCh38
rs771235755 Chr.11: 67483275-67483275 on Build GRCh38
rs200264737 Chr.11: 67483284-67483284 on Build GRCh38
rs989975901 Chr.11: 67483156-67483156 on Build GRCh38
rs773873262 Chr.11: 67483286-67483286 on Build GRCh38
rs765880091 Chr.11: 67483139-67483139 on Build GRCh38
rs116940576 Chr.11: 67483226-67483226 on Build GRCh38
rs760006823 Chr.11: 67483233-67483233 on Build GRCh38
rs267606584 Chr.11: 67483246-67483247 on Build GRCh38
rs770283272 Chr.11: 67483283-67483283 on Build GRCh38
rs199681649 Chr.11: 67483146-67483146 on Build GRCh38
rs901819028 Chr.11: 67483180-67483180 on Build GRCh38
rs121908357 Chr.11: 67483222-67483222 on Build GRCh38
rs199913396 Chr.11: 67483221-67483221 on Build GRCh38
rs554611101 Chr.11: 67483335-67483335 on Build GRCh38
rs201958318 Chr.11: 67483230-67483230 on Build GRCh38
rs773224238 Chr.11: 67483211-67483211 on Build GRCh38
rs145047094 Chr.11: 67483205-67483205 on Build GRCh38
rs768090582 Chr.11: 67483321-67483321 on Build GRCh38
rs139459091 Chr.11: 67483184-67483184 on Build GRCh38
rs763596431 Chr.11: 67483245-67483245 on Build GRCh38
rs374003812 Chr.11: 67483266-67483266 on Build GRCh38
rs376913545 Chr.11: 67483196-67483196 on Build GRCh38
rs747079315 Chr.11: 67483270-67483270 on Build GRCh38
rs755830428 Chr.11: 67483296-67483296 on Build GRCh38
rs549056286 Chr.11: 67483204-67483204 on Build GRCh38
rs777211577 Chr.11: 67483148-67483148 on Build GRCh38
rs756887504 Chr.11: 67483250-67483250 on Build GRCh38
rs776958604 Chr.11: 67483280-67483280 on Build GRCh38
rs745693426 Chr.11: 67483192-67483192 on Build GRCh38
rs750116890 Chr.11: 67483255-67483255 on Build GRCh38
rs373339489 Chr.11: 67483295-67483295 on Build GRCh38
rs551077555 Chr.11: 67483147-67483147 on Build GRCh38
rs756870384 Chr.11: 67483170-67483170 on Build GRCh38
rs1024903808 Chr.11: 67483237-67483237 on Build GRCh38
rs267606568 Chr.11: 67483228-67483228 on Build GRCh38
rs267606562 Chr.11: 67483154-67483154 on Build GRCh38
rs79662690 Chr.11: 67483194-67483194 on Build GRCh38
rs267606546 Chr.11: 67483160-67483160 on Build GRCh38
rs542623040 Chr.11: 67483306-67483306 on Build GRCh38
rs969013352 Chr.11: 67483243-67483243 on Build GRCh38
rs1015444550 Chr.11: 67483281-67483281 on Build GRCh38
rs1040608296 Chr.11: 67483144-67483144 on Build GRCh38
rs104894194 Chr.11: 67483198-67483198 on Build GRCh38
rs777083581 Chr.11: 67483231-67483231 on Build GRCh38
rs755881873 Chr.11: 67483257-67483257 on Build GRCh38
rs201382386 Chr.11: 67483282-67483282 on Build GRCh38
rs749243552 Chr.11: 67483265-67483265 on Build GRCh38
rs104895074 Chr.11: 67483232-67483239 on Build GRCh38
rs751208166 Chr.11: 67483153-67483153 on Build GRCh38
rs371636632 Chr.11: 67483251-67483251 on Build GRCh38
rs1028243090 Chr.11: 67483314-67483314 on Build GRCh38
rs754893214 Chr.11: 67483309-67483309 on Build GRCh38
rs377710724 Chr.11: 67483157-67483157 on Build GRCh38
rs267606567 Chr.11: 67483224-67483229 on Build GRCh38
rs764533249 Chr.11: 67483285-67483285 on Build GRCh38
rs777849377 Chr.11: 67483268-67483268 on Build GRCh38
rs954227691 Chr.11: 67483349-67483349 on Build GRCh38
rs747813691 Chr.11: 67483266-67483270 on Build GRCh38
rs187051018 Chr.11: 67483267-67483267 on Build GRCh38
rs573023826 Chr.11: 67483351-67483351 on Build GRCh38
rs555159979 Chr.11: 67483181-67483181 on Build GRCh38

View Full Product Details