Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 200644169-200644400 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX59
Entrez Gene ID
83479
Gene Name
DEAD-box helicase 59
Gene Aliases
OFD5, ZNHIT5
UniGene
Hs.497332
Gene Chromosome Location
Chr.1: 200640807-200669998 on Build GRCh38
Transcripts
NM_001031725.5 NM_001320181.1 NM_001320182.1 XM_005245519.1 XM_005245521.2 XM_006711562.1 XM_011510035.2 XM_017002431.1 XM_017002432.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1195277 c.1786C>G p.Q596E Chr.1: 200644328-200644328 on Build GRCh38
3671580 c.1736C>A p.P579Q Chr.1: 200644378-200644378 on Build GRCh38
1203289 c.1850C>A p.S617Y Chr.1: 200644264-200644264 on Build GRCh38
4026786 c.1767A>C p.Q589H Chr.1: 200644347-200644347 on Build GRCh38
2126297 c.1722G>A p.Q574Q Chr.1: 200644392-200644392 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751501368 Chr.1: 200644302-200644302 on Build GRCh38
rs201990304 Chr.1: 200644234-200644234 on Build GRCh38
rs748800077 Chr.1: 200644254-200644254 on Build GRCh38
rs750231186 Chr.1: 200644311-200644311 on Build GRCh38
rs772359313 Chr.1: 200644287-200644287 on Build GRCh38
rs904886878 Chr.1: 200644362-200644362 on Build GRCh38
rs759757683 Chr.1: 200644297-200644297 on Build GRCh38
rs745481478 Chr.1: 200644225-200644225 on Build GRCh38
rs779480651 Chr.1: 200644232-200644232 on Build GRCh38
rs777947658 Chr.1: 200644233-200644234 on Build GRCh38
rs769198531 Chr.1: 200644255-200644255 on Build GRCh38
rs753328906 Chr.1: 200644350-200644350 on Build GRCh38
rs775179019 Chr.1: 200644226-200644226 on Build GRCh38
rs779748662 Chr.1: 200644338-200644338 on Build GRCh38
rs771295468 Chr.1: 200644292-200644292 on Build GRCh38
rs147039616 Chr.1: 200644288-200644288 on Build GRCh38
rs748561193 Chr.1: 200644369-200644369 on Build GRCh38
rs754491646 Chr.1: 200644360-200644360 on Build GRCh38
rs755695604 Chr.1: 200644319-200644319 on Build GRCh38
rs757060233 Chr.1: 200644219-200644219 on Build GRCh38
rs1047894813 Chr.1: 200644243-200644243 on Build GRCh38
rs200630479 Chr.1: 200644225-200644225 on Build GRCh38
rs751365711 Chr.1: 200644213-200644213 on Build GRCh38
rs767166701 Chr.1: 200644309-200644309 on Build GRCh38
rs779440784 Chr.1: 200644365-200644365 on Build GRCh38
rs763943908 Chr.1: 200644212-200644212 on Build GRCh38
rs768840620 Chr.1: 200644252-200644252 on Build GRCh38
rs145775065 Chr.1: 200644264-200644264 on Build GRCh38
rs142642032 Chr.1: 200644291-200644291 on Build GRCh38
rs753184083 Chr.1: 200644224-200644225 on Build GRCh38
rs563763124 Chr.1: 200644221-200644221 on Build GRCh38
rs776788636 Chr.1: 200644294-200644294 on Build GRCh38
rs748521357 Chr.1: 200644285-200644285 on Build GRCh38
rs761639527 Chr.1: 200644307-200644307 on Build GRCh38

View Full Product Details