Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81671951-81672224 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC137
Entrez Gene ID
339230
Gene Name
coiled-coil domain containing 137
Gene Aliases
RaRF
UniGene
Hs.744125
Gene Chromosome Location
Chr.17: 81666697-81673907 on Build GRCh38
Transcripts
NM_199287.2 XM_011524738.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2805398 c.596G>T p.G199V Chr.17: 81672091-81672091 on Build GRCh38
6026007 c.645C>T p.S215S Chr.17: 81672140-81672140 on Build GRCh38
4922337 c.660G>A p.Q220Q Chr.17: 81672155-81672155 on Build GRCh38
985898 c.621G>T p.E207D Chr.17: 81672116-81672116 on Build GRCh38
3523667 c.636C>T p.P212P Chr.17: 81672131-81672131 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779772538 Chr.17: 81672180-81672180 on Build GRCh38
rs758736888 Chr.17: 81672163-81672168 on Build GRCh38
rs1035863759 Chr.17: 81672132-81672132 on Build GRCh38
rs373936708 Chr.17: 81672112-81672112 on Build GRCh38
rs745696787 Chr.17: 81672116-81672116 on Build GRCh38
rs767994197 Chr.17: 81672079-81672079 on Build GRCh38
rs376603100 Chr.17: 81672089-81672089 on Build GRCh38
rs779482910 Chr.17: 81672107-81672108 on Build GRCh38
rs780174190 Chr.17: 81672183-81672183 on Build GRCh38
rs1012820767 Chr.17: 81672028-81672028 on Build GRCh38
rs1017587760 Chr.17: 81672156-81672156 on Build GRCh38
rs768758231 Chr.17: 81672191-81672191 on Build GRCh38
rs552243530 Chr.17: 81672150-81672150 on Build GRCh38
rs201200598 Chr.17: 81672090-81672090 on Build GRCh38
rs777319385 Chr.17: 81672106-81672106 on Build GRCh38
rs759977481 Chr.17: 81672036-81672036 on Build GRCh38
rs200346416 Chr.17: 81672060-81672060 on Build GRCh38
rs192264445 Chr.17: 81672047-81672047 on Build GRCh38
rs764346327 Chr.17: 81672055-81672055 on Build GRCh38
rs183627821 Chr.17: 81672115-81672115 on Build GRCh38
rs780635957 Chr.17: 81672096-81672096 on Build GRCh38
rs745583974 Chr.17: 81672177-81672177 on Build GRCh38
rs755952019 Chr.17: 81672178-81672178 on Build GRCh38
rs746215609 Chr.17: 81672159-81672160 on Build GRCh38
rs891819943 Chr.17: 81672067-81672067 on Build GRCh38
rs763070699 Chr.17: 81672129-81672129 on Build GRCh38
rs201286650 Chr.17: 81672169-81672169 on Build GRCh38
rs201656915 Chr.17: 81672141-81672141 on Build GRCh38
rs763158648 Chr.17: 81672051-81672051 on Build GRCh38
rs775380947 Chr.17: 81672041-81672041 on Build GRCh38
rs755542085 Chr.17: 81672153-81672153 on Build GRCh38
rs12940151 Chr.17: 81672015-81672015 on Build GRCh38
rs771519473 Chr.17: 81671983-81671983 on Build GRCh38
rs775504743 Chr.17: 81672128-81672128 on Build GRCh38
rs7502870 Chr.17: 81672168-81672168 on Build GRCh38
rs1056698539 Chr.17: 81671987-81671987 on Build GRCh38
rs769818742 Chr.17: 81672040-81672040 on Build GRCh38
rs912430997 Chr.17: 81671990-81671990 on Build GRCh38
rs373550389 Chr.17: 81672155-81672155 on Build GRCh38
rs200887312 Chr.17: 81672033-81672033 on Build GRCh38
rs761134731 Chr.17: 81672149-81672149 on Build GRCh38
rs762184231 Chr.17: 81672069-81672069 on Build GRCh38
rs376019013 Chr.17: 81672140-81672140 on Build GRCh38
rs1026159450 Chr.17: 81672114-81672114 on Build GRCh38
rs757915022 Chr.17: 81672105-81672105 on Build GRCh38
rs768844136 Chr.17: 81672139-81672139 on Build GRCh38
rs749233063 Chr.17: 81672190-81672190 on Build GRCh38
rs752078772 Chr.17: 81672102-81672102 on Build GRCh38
rs770610858 Chr.17: 81672113-81672113 on Build GRCh38
rs751117971 Chr.17: 81672160-81672160 on Build GRCh38

View Full Product Details