Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 25158388-25158625 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C7orf31
Entrez Gene ID
136895
Gene Name
chromosome 7 open reading frame 31
Gene Aliases
-
UniGene
Hs.122055
Gene Chromosome Location
Chr.7: 25134697-25180748 on Build GRCh38
Transcripts
NM_138811.3 XM_005249619.3 XM_005249620.3 XM_011515125.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1088738 c.388C>T p.P130S Chr.7: 25158526-25158526 on Build GRCh38
5798786 c.409G>C p.V137L Chr.7: 25158505-25158505 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs140027256 Chr.7: 25158507-25158507 on Build GRCh38
rs373873012 Chr.7: 25158429-25158429 on Build GRCh38
rs1019343735 Chr.7: 25158448-25158448 on Build GRCh38
rs753148852 Chr.7: 25158502-25158502 on Build GRCh38
rs760388646 Chr.7: 25158563-25158563 on Build GRCh38
rs191500428 Chr.7: 25158519-25158519 on Build GRCh38
rs369438213 Chr.7: 25158489-25158489 on Build GRCh38
rs903967337 Chr.7: 25158412-25158412 on Build GRCh38
rs375072377 Chr.7: 25158484-25158484 on Build GRCh38
rs768223079 Chr.7: 25158458-25158458 on Build GRCh38
rs982497251 Chr.7: 25158549-25158549 on Build GRCh38
rs771644492 Chr.7: 25158554-25158554 on Build GRCh38
rs1003891513 Chr.7: 25158436-25158436 on Build GRCh38
rs374948610 Chr.7: 25158546-25158546 on Build GRCh38
rs761550694 Chr.7: 25158466-25158466 on Build GRCh38
rs773982864 Chr.7: 25158550-25158550 on Build GRCh38
rs781306808 Chr.7: 25158525-25158525 on Build GRCh38
rs747702925 Chr.7: 25158553-25158553 on Build GRCh38
rs545407521 Chr.7: 25158447-25158447 on Build GRCh38
rs755812925 Chr.7: 25158534-25158534 on Build GRCh38
rs765656540 Chr.7: 25158493-25158493 on Build GRCh38
rs202153993 Chr.7: 25158457-25158457 on Build GRCh38
rs746129970 Chr.7: 25158532-25158532 on Build GRCh38
rs2523066 Chr.7: 25158470-25158470 on Build GRCh38
rs749169631 Chr.7: 25158538-25158538 on Build GRCh38
rs774000729 Chr.7: 25158459-25158459 on Build GRCh38
rs377045223 Chr.7: 25158566-25158566 on Build GRCh38
rs977479676 Chr.7: 25158526-25158526 on Build GRCh38
rs990808240 Chr.7: 25158496-25158496 on Build GRCh38
rs775903181 Chr.7: 25158581-25158581 on Build GRCh38
rs143559120 Chr.7: 25158518-25158518 on Build GRCh38
rs113525450 Chr.7: 25158449-25158449 on Build GRCh38
rs955361498 Chr.7: 25158448-25158449 on Build GRCh38
rs145645287 Chr.7: 25158562-25158562 on Build GRCh38

View Full Product Details