Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 34348121-34348390 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUTM1
Entrez Gene ID
256646
Gene Name
NUT midline carcinoma family member 1
Gene Aliases
C15orf55, FAM22H, NUT
UniGene
Hs.525769
Gene Chromosome Location
Chr.15: 34343315-34357737 on Build GRCh38
Transcripts
NM_001284292.1 NM_001284293.1 NM_175741.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1198150 c.309A>C p.Q103H Chr.15: 34348261-34348261 on Build GRCh38
6849332 c.298G>T p.E100* Chr.15: 34348250-34348250 on Build GRCh38
960838 c.313C>T p.Q105* Chr.15: 34348265-34348265 on Build GRCh38
960837 c.279G>T p.K93N Chr.15: 34348231-34348231 on Build GRCh38
4545281 c.374G>A p.G125D Chr.15: 34348326-34348326 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs188615920 Chr.15: 34348340-34348340 on Build GRCh38
rs752190210 Chr.15: 34348289-34348289 on Build GRCh38
rs576817548 Chr.15: 34348298-34348298 on Build GRCh38
rs750888502 Chr.15: 34348153-34348153 on Build GRCh38
rs535995310 Chr.15: 34348178-34348178 on Build GRCh38
rs749054182 Chr.15: 34348213-34348213 on Build GRCh38
rs765632741 Chr.15: 34348342-34348342 on Build GRCh38
rs745754441 Chr.15: 34348230-34348230 on Build GRCh38
rs554363938 Chr.15: 34348233-34348233 on Build GRCh38
rs1028971027 Chr.15: 34348172-34348177 on Build GRCh38
rs376023605 Chr.15: 34348362-34348362 on Build GRCh38
rs931115519 Chr.15: 34348253-34348253 on Build GRCh38
rs776132811 Chr.15: 34348202-34348202 on Build GRCh38
rs147877754 Chr.15: 34348261-34348261 on Build GRCh38
rs779291598 Chr.15: 34348149-34348149 on Build GRCh38
rs773563082 Chr.15: 34348343-34348343 on Build GRCh38
rs767081750 Chr.15: 34348283-34348283 on Build GRCh38
rs373216708 Chr.15: 34348360-34348360 on Build GRCh38
rs1042643515 Chr.15: 34348241-34348241 on Build GRCh38
rs765586587 Chr.15: 34348259-34348259 on Build GRCh38
rs141502234 Chr.15: 34348303-34348303 on Build GRCh38
rs370488235 Chr.15: 34348317-34348317 on Build GRCh38
rs906531581 Chr.15: 34348316-34348316 on Build GRCh38
rs972575412 Chr.15: 34348242-34348242 on Build GRCh38
rs574609632 Chr.15: 34348327-34348327 on Build GRCh38
rs780660815 Chr.15: 34348162-34348162 on Build GRCh38
rs758873419 Chr.15: 34348281-34348281 on Build GRCh38
rs770696098 Chr.15: 34348214-34348214 on Build GRCh38
rs949082934 Chr.15: 34348312-34348312 on Build GRCh38
rs745491067 Chr.15: 34348319-34348319 on Build GRCh38
rs932399842 Chr.15: 34348168-34348168 on Build GRCh38
rs1015904080 Chr.15: 34348337-34348337 on Build GRCh38
rs200731305 Chr.15: 34348239-34348239 on Build GRCh38
rs776846562 Chr.15: 34348251-34348251 on Build GRCh38
rs202160501 Chr.15: 34348238-34348238 on Build GRCh38
rs961440239 Chr.15: 34348227-34348227 on Build GRCh38
rs370054299 Chr.15: 34348142-34348142 on Build GRCh38
rs774464902 Chr.15: 34348220-34348220 on Build GRCh38
rs777299530 Chr.15: 34348296-34348296 on Build GRCh38
rs762115937 Chr.15: 34348254-34348254 on Build GRCh38
rs893405516 Chr.15: 34348152-34348152 on Build GRCh38
rs199906242 Chr.15: 34348294-34348294 on Build GRCh38
rs199644358 Chr.15: 34348299-34348299 on Build GRCh38
rs764232519 Chr.15: 34348246-34348246 on Build GRCh38
rs377416887 Chr.15: 34348308-34348308 on Build GRCh38
rs778831851 Chr.15: 34348300-34348300 on Build GRCh38
rs777681816 Chr.15: 34348183-34348183 on Build GRCh38
rs543959983 Chr.15: 34348309-34348309 on Build GRCh38
rs754504740 Chr.15: 34348161-34348161 on Build GRCh38
rs912221144 Chr.15: 34348196-34348196 on Build GRCh38
rs768343381 Chr.15: 34348171-34348172 on Build GRCh38
rs936805048 Chr.15: 34348271-34348271 on Build GRCh38
rs761012005 Chr.15: 34348245-34348245 on Build GRCh38
rs430083 Chr.15: 34348177-34348177 on Build GRCh38

View Full Product Details