Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 112039043-112039292 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLAT
Entrez Gene ID
1737
Gene Name
dihydrolipoamide S-acetyltransferase
Gene Aliases
DLTA, PDC-E2, PDCE2
UniGene
Hs.335551
Gene Chromosome Location
Chr.11: 112024814-112064278 on Build GRCh38
Transcripts
NM_001931.4 XM_011542647.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
348334 c.979G>T p.A327S Chr.11: 112039247-112039247 on Build GRCh38
6644519 c.995C>T p.T332I Chr.11: 112039263-112039263 on Build GRCh38
3415535 c.982G>A p.A328T Chr.11: 112039250-112039250 on Build GRCh38
250147 c.1002G>A p.Q334Q Chr.11: 112039270-112039270 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs782800078 Chr.11: 112039229-112039230 on Build GRCh38
rs587736661 Chr.11: 112039265-112039265 on Build GRCh38
rs139204615 Chr.11: 112039129-112039129 on Build GRCh38
rs587593887 Chr.11: 112039173-112039173 on Build GRCh38
rs78320677 Chr.11: 112039259-112039259 on Build GRCh38
rs947504064 Chr.11: 112039146-112039146 on Build GRCh38
rs914672141 Chr.11: 112039136-112039136 on Build GRCh38
rs367875541 Chr.11: 112039243-112039243 on Build GRCh38
rs886090417 Chr.11: 112039231-112039231 on Build GRCh38
rs782139204 Chr.11: 112039211-112039211 on Build GRCh38
rs781861956 Chr.11: 112039251-112039251 on Build GRCh38
rs900707924 Chr.11: 112039156-112039156 on Build GRCh38
rs782667970 Chr.11: 112039260-112039260 on Build GRCh38
rs781853215 Chr.11: 112039239-112039239 on Build GRCh38
rs782701797 Chr.11: 112039237-112039237 on Build GRCh38
rs76539854 Chr.11: 112039186-112039186 on Build GRCh38
rs782024604 Chr.11: 112039202-112039202 on Build GRCh38
rs782478109 Chr.11: 112039247-112039247 on Build GRCh38
rs782763586 Chr.11: 112039199-112039199 on Build GRCh38
rs200611949 Chr.11: 112039155-112039155 on Build GRCh38
rs782453793 Chr.11: 112039266-112039266 on Build GRCh38
rs6589260 Chr.11: 112039095-112039095 on Build GRCh38
rs782157762 Chr.11: 112039249-112039249 on Build GRCh38

View Full Product Details