Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 17398260-17398519 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCC8
Entrez Gene ID
6833
Gene Name
ATP binding cassette subfamily C member 8
Gene Aliases
ABC36, HHF1, HI, HRINS, MRP8, PHHI, SUR, SUR1, SUR1delta2, TNDM2
UniGene
Hs.54470
Gene Chromosome Location
Chr.11: 17392885-17476870 on Build GRCh38
Transcripts
NM_000352.4 NM_001287174.1 XM_011520331.2 XM_017018197.1 XM_017018198.1 XM_017018199.1 XM_017018200.1 XM_017018201.1 XM_017018202.1 XM_017018204.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3868896 c.3753G>A p.M1251I Chr.11: 17398339-17398339 on Build GRCh38
6315418 c.3725C>A p.A1242D Chr.11: 17398367-17398367 on Build GRCh38
6014306 c.3714C>T p.L1238L Chr.11: 17398378-17398378 on Build GRCh38
3445817 c.3702C>T p.N1234N Chr.11: 17398390-17398390 on Build GRCh38
6131952 c.3712C>T p.L1238F Chr.11: 17398380-17398380 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1057516281 Chr.11: 17398344-17398344 on Build GRCh38
rs760117593 Chr.11: 17398475-17398475 on Build GRCh38
rs975856232 Chr.11: 17398343-17398343 on Build GRCh38
rs759280196 Chr.11: 17398462-17398462 on Build GRCh38
rs761599545 Chr.11: 17398488-17398488 on Build GRCh38
rs768820409 Chr.11: 17398442-17398442 on Build GRCh38
rs776842732 Chr.11: 17398313-17398313 on Build GRCh38
rs779489003 Chr.11: 17398378-17398378 on Build GRCh38
rs772031866 Chr.11: 17398473-17398473 on Build GRCh38
rs953395303 Chr.11: 17398396-17398396 on Build GRCh38
rs775759502 Chr.11: 17398290-17398290 on Build GRCh38
rs17846743 Chr.11: 17398312-17398312 on Build GRCh38
rs759513178 Chr.11: 17398328-17398328 on Build GRCh38
rs763659404 Chr.11: 17398360-17398360 on Build GRCh38
rs374298515 Chr.11: 17398430-17398430 on Build GRCh38
rs758995284 Chr.11: 17398410-17398410 on Build GRCh38
rs1046824013 Chr.11: 17398293-17398293 on Build GRCh38
rs753284772 Chr.11: 17398482-17398482 on Build GRCh38
rs774183901 Chr.11: 17398447-17398447 on Build GRCh38
rs886048049 Chr.11: 17398387-17398387 on Build GRCh38
rs763187143 Chr.11: 17398298-17398298 on Build GRCh38
rs753473058 Chr.11: 17398363-17398363 on Build GRCh38
rs770768484 Chr.11: 17398318-17398318 on Build GRCh38
rs747063115 Chr.11: 17398417-17398417 on Build GRCh38
rs774124906 Chr.11: 17398326-17398326 on Build GRCh38
rs376023733 Chr.11: 17398298-17398298 on Build GRCh38
rs567382793 Chr.11: 17398431-17398431 on Build GRCh38
rs769968875 Chr.11: 17398438-17398438 on Build GRCh38
rs768706912 Chr.11: 17398410-17398410 on Build GRCh38
rs767422948 Chr.11: 17398334-17398334 on Build GRCh38
rs760196276 Chr.11: 17398337-17398337 on Build GRCh38
rs762665556 Chr.11: 17398315-17398315 on Build GRCh38
rs370636988 Chr.11: 17398411-17398411 on Build GRCh38
rs756849559 Chr.11: 17398367-17398367 on Build GRCh38
rs763718966 Chr.11: 17398480-17398480 on Build GRCh38
rs750793918 Chr.11: 17398406-17398406 on Build GRCh38
rs923215419 Chr.11: 17398392-17398392 on Build GRCh38
rs748466485 Chr.11: 17398435-17398435 on Build GRCh38

View Full Product Details