Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 25835631-25835892 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AUNIP
Entrez Gene ID
79000
Gene Name
aurora kinase A and ninein interacting protein
Gene Aliases
AIBP, C1orf135
UniGene
Hs.149305
Gene Chromosome Location
Chr.1: 25831913-25859458 on Build GRCh38
Transcripts
NM_001287490.1 NM_024037.2 XM_017002300.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4030906 c.353C>A p.P118H Chr.1: 25835714-25835714 on Build GRCh38
3773490 c.353C>T p.P118L Chr.1: 25835714-25835714 on Build GRCh38
7195035 c.300G>C p.A100A Chr.1: 25835767-25835767 on Build GRCh38
7282094 c.270T>A p.S90R Chr.1: 25835797-25835797 on Build GRCh38
6608834 c.328T>C p.L110L Chr.1: 25835739-25835739 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768357310 Chr.1: 25835798-25835798 on Build GRCh38
rs755159717 Chr.1: 25835751-25835751 on Build GRCh38
rs762152604 Chr.1: 25835724-25835724 on Build GRCh38
rs762205571 Chr.1: 25835804-25835804 on Build GRCh38
rs749444706 Chr.1: 25835785-25835786 on Build GRCh38
rs777673955 Chr.1: 25835652-25835652 on Build GRCh38
rs1044703448 Chr.1: 25835853-25835853 on Build GRCh38
rs764373565 Chr.1: 25835719-25835719 on Build GRCh38
rs910381318 Chr.1: 25835761-25835761 on Build GRCh38
rs201139324 Chr.1: 25835821-25835821 on Build GRCh38
rs200152788 Chr.1: 25835682-25835682 on Build GRCh38
rs751763350 Chr.1: 25835744-25835744 on Build GRCh38
rs547863335 Chr.1: 25835692-25835692 on Build GRCh38
rs377641827 Chr.1: 25835731-25835731 on Build GRCh38
rs745794955 Chr.1: 25835777-25835777 on Build GRCh38
rs777721119 Chr.1: 25835757-25835757 on Build GRCh38
rs867761704 Chr.1: 25835720-25835720 on Build GRCh38
rs145647141 Chr.1: 25835660-25835660 on Build GRCh38
rs550451631 Chr.1: 25835671-25835671 on Build GRCh38
rs375324621 Chr.1: 25835818-25835818 on Build GRCh38
rs201314032 Chr.1: 25835667-25835667 on Build GRCh38
rs776261811 Chr.1: 25835801-25835801 on Build GRCh38
rs754995601 Chr.1: 25835834-25835834 on Build GRCh38
rs34449716 Chr.1: 25835822-25835822 on Build GRCh38
rs766436655 Chr.1: 25835742-25835742 on Build GRCh38
rs777711727 Chr.1: 25835704-25835705 on Build GRCh38
rs767914475 Chr.1: 25835836-25835836 on Build GRCh38
rs769501757 Chr.1: 25835651-25835651 on Build GRCh38
rs111227957 Chr.1: 25835850-25835850 on Build GRCh38
rs779034171 Chr.1: 25835771-25835771 on Build GRCh38
rs141597954 Chr.1: 25835768-25835768 on Build GRCh38
rs148257460 Chr.1: 25835796-25835796 on Build GRCh38
rs767690932 Chr.1: 25835786-25835786 on Build GRCh38
rs771502890 Chr.1: 25835778-25835778 on Build GRCh38
rs200879396 Chr.1: 25835669-25835669 on Build GRCh38
rs748057926 Chr.1: 25835667-25835670 on Build GRCh38
rs765800562 Chr.1: 25835730-25835730 on Build GRCh38
rs756087178 Chr.1: 25835667-25835668 on Build GRCh38
rs772304245 Chr.1: 25835664-25835664 on Build GRCh38
rs758460528 Chr.1: 25835733-25835733 on Build GRCh38
rs576640245 Chr.1: 25835812-25835812 on Build GRCh38
rs570224400 Chr.1: 25835676-25835676 on Build GRCh38
rs202007540 Chr.1: 25835767-25835767 on Build GRCh38
rs776924890 Chr.1: 25835723-25835723 on Build GRCh38
rs765567579 Chr.1: 25835810-25835810 on Build GRCh38

View Full Product Details